Skip to Content
Merck
All Photos(1)

Key Documents

19-3760

Sigma-Aldrich

Poly(methyl methacrylate)

CP

Synonym(s):

PMMA, Poly(methacrylic acid methyl ester)

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
[CH2C(CH3)(CO2CH3)]n
CAS Number:
MDL number:
UNSPSC Code:
41116107

grade

CP

autoignition temp.

580 °F

availability

available only in Japan

InChI

1S/C5H9O2/c1-4(2)5(6)7-3/h1-3H3

InChI key

PMAMJWJDBDSDHV-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Storage Class Code

11 - Combustible Solids

WGK

nwg

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

S A Bini et al.
The bone & joint journal, 95-B(3), 367-370 (2013-03-02)
We evaluated the impact of pre-coating the tibial component with polymethylmethacrylate (PMMA) on implant survival in a cohort of 16 548 primary NexGen total knee replacements (TKRs) in 14 113 patients. In 13 835 TKRs a pre-coated tray was used
Kariem Mostafa Al Batouty et al.
The journal of adhesive dentistry, 15(2), 161-166 (2013-03-28)
To compare the push-out bond strength of a new glass fiber root canal filling material to that of Resilon/ Epiphany, EndoREZ, and gutta-percha/AH Plus root canal filling materials. Sixty roots were randomly divided into four groups (n = 15) according
Pleural infection 55 years after plombage for pulmonary tuberculosis.
Qian-kun Chen et al.
Asian cardiovascular & thoracic annals, 21(1), 100-100 (2013-02-23)
Gursukhman S Sidhu et al.
Journal of neurosurgery. Spine, 19(1), 61-70 (2013-05-07)
The authors endeavor to highlight the surgical management of severe neurological deficit resulting from cement leakage after percutaneous vertebroplasty and to systematically review the literature on the management of this complication. A patient presented after a vertebroplasty procedure for traumatic
Distortions of iris and anterior lens capsule OCT images by ICRS.
Hosik Hwang et al.
Journal of refractive surgery (Thorofare, N.J. : 1995), 29(3), 155-156 (2013-03-01)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service