Skip to Content
Merck
All Photos(3)

Key Documents

182230

Sigma-Aldrich

Poly(methyl methacrylate)

Synonym(s):

PMMA, Poly(methacrylic acid methyl ester)

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
[CH2C(CH3)(CO2CH3)]n
CAS Number:
MDL number:
UNSPSC Code:
12162002
PubChem Substance ID:
NACRES:
NA.23

autoignition temp.

580 °F

impurities

may contain ≤2.0% toluene

refractive index

n20/D 1.49

inherent viscosity

0.20 dL/g(lit.)

transition temp

Tg (DSC) 105 °C (midpoint)

solubility

H2O: insoluble

density

1.188 g/mL at 25 °C

InChI

1S/C5H9O2/c1-4(2)5(6)7-3/h1-3H3

InChI key

PMAMJWJDBDSDHV-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Poly (methyl methacrylate) (PMMA) is an amorphous transparent thermoplastic polymer. PMMA is recognized as an optical polymer based on its refractive index of 1.49. Hence, it is used in optical fibers.It finds uses in biological applications because of its lower water absorption capability and biocompatible.The maximum moisture content of PMMA is 1.71% and SBF absorption is 1.96%.Simulated Body fluid test (SBF) is a method to characterize the in vitro bioactivity of ceramic materials, by immersing the materials in an aqueous SBF solution.

Application

PMMA/HA (Hydroxyapatite) composites have been used in biomedical applications such as dentistry, orthopedic retainers, and bone replacement. It is used as substrate for graphene growth.
Some studies report its use in
PMMA-titania hybrid optical thins films
PMMA/polystryrene/clay nanocomposites
PMMA/polyurethane/carbon black nanocomposites for methanol fuel cells

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Christian J F Bertens et al.
European journal of pharmaceutics and biopharmaceutics : official journal of Arbeitsgemeinschaft fur Pharmazeutische Verfahrenstechnik e.V, 150, 120-130 (2020-03-17)
Eye drops and ointments are the most prescribed methods for ocular drug delivery. However, due to low drug bioavailability, rapid drug elimination, and low patient compliance there is a need for improved ophthalmic drug delivery systems. This study provides insights
Synthesis of Ethylene Glycol Dimethacrylate-Methyl Methacrylate Copolymers, Determination of their Reactivity Ratios, and a Study of Dopant and Temperature Effects on their Conductivities
Ramelow US and Pingili S
Polymer, 2(3), 265-285 (2010)
Poly (methyl methacrylate) and polystyrene/clay nanocomposites prepared by in-situ polymerization.
Zeng C and Lee LJ
Macromolecules, 34(12), 4098-4103 (2001)
Jian-Chiun Liou et al.
Nanoscale research letters, 9(1), 1-1 (2014-01-02)
In this study, the Mo-electrode thin films were deposited by a two-stepped process, and the high-purity copper indium selenide-based powder (CuInSe2, CIS) was fabricated by hydrothermal process by Nanowin Technology Co. Ltd. From the X-ray pattern of the CIS precursor
High-refractive-index thin films prepared from trialkoxysilane-capped poly (methyl methacrylate)-titania materials
Lee LH, et al.
Chemistry of Materials, 13(3), 1137-1142 (2001)

Articles

The detection and quantification of biomarkers are essential for medical diagnostics, environmental monitoring, and bioresearch.

Electrospinning technique applications discussed, emphasizing control of nanofibers and assembly into 3D architectures.

Electrospinning technique applications discussed, emphasizing control of nanofibers and assembly into 3D architectures.

Electrospinning technique applications discussed, emphasizing control of nanofibers and assembly into 3D architectures.

See All

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service