Skip to Content
Merck
All Photos(1)

Key Documents

19-3550

Sigma-Aldrich

4-Methyl-2-pentanone

SAJ first grade, ≥99.0%

Synonym(s):

Isobutyl methyl ketone, Isopropylacetone, MIBK, Methyl isobutyl ketone

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3)2CHCH2COCH3
CAS Number:
Molecular Weight:
100.16
Beilstein:
605399
MDL number:
UNSPSC Code:
12352115
PubChem Substance ID:

grade

SAJ first grade

vapor density

3.5 (vs air)

vapor pressure

15.7 mmHg

Assay

≥99.0%

form

liquid

autoignition temp.

840 °F

expl. lim.

1.2-8 %, 93 °F

availability

available only in Japan

refractive index

n20/D 1.395 (lit.)

bp

117-118 °C

mp

−80 °C (lit.)

density

0.801 g/mL at 25 °C (lit.)

SMILES string

CC(C)CC(C)=O

InChI

1S/C6H12O/c1-5(2)4-6(3)7/h5H,4H2,1-3H3

InChI key

NTIZESTWPVYFNL-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Inhalation - Carc. 2 Inhalation - Eye Irrit. 2 - Flam. Liq. 2 - STOT SE 3

Target Organs

Central nervous system

Supplementary Hazards

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

57.2 °F

Flash Point(C)

14 °C


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

H J Shim et al.
Journal of chromatography. B, Biomedical applications, 656(2), 407-414 (1994-06-17)
A high-performance liquid chromatographic method was developed for the simultaneous determination of a new anthracycline, DA-125 (I), and its metabolites (M1, M2, M3, and M4) in rat plasma and urine using fluorescein as an internal standard. Compound I, a prodrug
Mao Ting et al.
The Science of the total environment, 390(1), 97-108 (2007-11-03)
Volatile organic compounds (VOCs) in the air of Beijing City were measured at the heights of 8, 32, 140 and 280 m on the Beijing 325 m meteorological tower in autumn 2005. Concentrations of fifty-five compounds were determined by quantitative
David Quesnel et al.
Water research, 39(4), 677-687 (2005-02-15)
An industrial wastewater that was pretreated by an aerobic thermophilic bacterial consortium (THE) was subjected to additional treatability studies by granular activated carbon (GAC) and a conventional activated sludge (CAS). The removal of dissolved organic carbon (DOC) in both systems
Ting-Ke Tseng et al.
Journal of hazardous materials, 122(1-2), 155-160 (2005-05-26)
Catalytic oxidation is one of the cost-effective technologies to solve the troublesome volatile organic compounds. This study treated methyl-isobutyl-ketone (MIBK) by a commercial catalyst, Pt/gamma-Al(2)O(3), in a fixed-bed reactor. The effects of operating factors, such as operating temperature, MIBK concentration
Daekeun Kim et al.
Chemosphere, 64(8), 1362-1368 (2006-02-10)
Single and ternary solute gas phase adsorption isotherms were conducted in this study to evaluate the effectiveness of a simple constant volume method, which was utilized by using Tedlar gas sampling bags as a constant volume batch reactor. For this

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service