Skip to Content
Merck
All Photos(1)

Key Documents

19-3500

Sigma-Aldrich

Iodomethane

SAJ special grade, ≥99.5%

Synonym(s):

Methyl iodide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
CH3I
CAS Number:
Molecular Weight:
141.94
Beilstein:
969135
MDL number:
UNSPSC Code:
12352101
PubChem Substance ID:
grade:
SAJ special grade
bp:
41-43 °C (lit.)
vapor pressure:
24.09 psi ( 55 °C)
7.89 psi ( 20 °C)

grade

SAJ special grade

vapor density

4.89 (vs air)

vapor pressure

24.09 psi ( 55 °C)
7.89 psi ( 20 °C)

Assay

≥99.5%

form

liquid

availability

available only in Japan

dilution

(for analytical testing)

refractive index

n20/D 1.531 (lit.)

bp

41-43 °C (lit.)

mp

−64 (lit.)

density

2.28 g/mL at 25 °C (lit.)

SMILES string

CI

InChI

1S/CH3I/c1-2/h1H3

InChI key

INQOMBQAUSQDDS-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Acute Tox. 4 Dermal - Aquatic Acute 1 - Aquatic Chronic 2 - Carc. 2 - Eye Dam. 1 - Flam. Liq. 3 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

89.8 °F - closed cup

Flash Point(C)

32.1 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

R Otto et al.
Nature chemistry, 4(7), 534-538 (2012-06-22)
Solvents have a profound influence on chemical reactions in solution and have long been used to control their outcome. Such effects are generally considered to be governed by thermodynamics; however, little is known about the steric effects of solvent molecules.
Microhydration effects on the intermediates of the S(N)2 reaction of iodide anion with methyl iodide.
Keisuke Doi et al.
Angewandte Chemie (International ed. in English), 52(16), 4380-4383 (2013-01-31)
Riet Bammens et al.
Glycobiology, 25(6), 669-682 (2015-01-23)
The congenital disorders of glycosylation (CDG), a group of inherited diseases characterized by aberrant glycosylation, encompass a wide range of defects, including glycosyltransferases, glycosidases, nucleotide-sugar transporters as well as proteins involved in maintaining Golgi architecture, pH and vesicular trafficking. Mutations
Warayuth Sajomsang et al.
International journal of biological macromolecules, 50(1), 263-269 (2011-11-22)
Five water-soluble chitosan derivatives were carried out by quaternizing either iodomethane or N-(3-chloro-2-hydroxypropyl) trimethylammonium chloride (Quat188) as a quaternizing agent under basic condition. The degree of quaternization (DQ) ranged between 28±2% and 90±2%. The antifungal activity was evaluated by using
Kazunori Kawamura et al.
Nuclear medicine and biology, 39(1), 89-99 (2011-08-13)
To explore the possible use of positron emission tomography (PET) probes for imaging of I(2)-imidazoline receptors (I(2)Rs) in peripheral tissues, we labeled two new I(2)R ligands, 2-[2-(o-tolyl)vinyl]-4,5-dihydro-1H-imidazole (K(i) for I(2)Rs, 3.7 nM) and 2-[2-(o-tolyl)ethyl]-4,5-dihydro-1H-imidazole (K(i) for I(2)Rs, 1.7 nM) with

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service