Skip to Content
Merck
All Photos(1)

Key Documents

07-4170

Sigma-Aldrich

2-Dimethylaminoethanol

SAJ first grade, ≥99.0%

Synonym(s):

N,N-Dimethyl-2-hydroxyethylamine, N,N-Dimethylethanolamine, Jeffcat DMEA

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3)2NCH2CH2OH
CAS Number:
Molecular Weight:
89.14
Beilstein:
1209235
EC Number:
MDL number:
UNSPSC Code:
12352104
PubChem Substance ID:

grade

SAJ first grade

vapor pressure

100 mmHg ( 55 °C)
6.12 mmHg ( 20 °C)

Assay

≥99.0%

availability

available only in Japan

refractive index

n20/D 1.4294 (lit.)

bp

134-136 °C (lit.)

mp

−70 °C (lit.)

density

0.886 g/mL at 20 °C (lit.)

SMILES string

CN(C)CCO

InChI

1S/C4H11NO/c1-5(2)3-4-6/h6H,3-4H2,1-2H3

InChI key

UEEJHVSXFDXPFK-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Inhalation - Acute Tox. 4 Dermal - Acute Tox. 4 Oral - Eye Dam. 1 - Flam. Liq. 3 - Skin Corr. 1B - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

104.0 °F - Seta closed cup

Flash Point(C)

40 °C - Seta closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

B Sommerfeld
Phytomedicine : international journal of phytotherapy and phytopharmacology, 14(11), 711-715 (2007-10-10)
Tricutan is a combination product of herbal extracts traditionally used for treatment of skin conditions, together with dimethylaminoethanol. The effectiveness of Tricutan in improving skin firmness and elasticity in photoaged facial skin was examined in a randomised, placebo-controlled, double-blind, split-face
Tadashi Tatsumi et al.
Bioorganic & medicinal chemistry, 18(7), 2720-2727 (2010-03-17)
Effects of retro-inverso (RI) modifications of HTLV-1 protease inhibitors containing a hydroxyethylamine isoster backbone were clarified. Construction of the isoster backbone was achieved by a stereoselective aldol reaction. Four diastereomers with different configurations at the isoster hydroxyl site and the
K M Houston et al.
Parasitology, 135(Pt 1), 55-61 (2007-09-26)
Juvenile female Litomosoides sigmodontis secrete a protein (Juv-p120) highly modified with dimethylethanolamine (DMAE). In an attempt to establish the source of this decoration worms were pulsed with [3H]-choline and [3H]-ethanolamine and the radio-isotope labelled products analysed. Both isotope labels were
Ouassila Amiri-Attou et al.
Molecules (Basel, Switzerland), 10(3), 545-551 (2007-11-17)
We report herein the synthesis of substituted 2-(6-nitrobenzo[1,3]dioxol-5-yl)-1- aryl ethanols and 2-(6-nitrobenzo[1,3]dioxol-5-yl)-propionic acid ethyl esters from the reaction of 5-chloromethyl-6-nitrobenzo[1,3]dioxole with various aromatic carbonyl and alpha- carbonyl ester derivatives using the tetrakis(dimethylamino)ethylene (TDAE) methodology.
Zhifeng Fu et al.
Analytica chimica acta, 638(2), 220-224 (2009-03-31)
Numerous drugs are carboxylic acid derivatives containing amino group, and hydrolysis reaction of these agents often generates toxic amines. Thus, the detection of amine impurity is of great importance in drug quality control of these amino group-containing ester and amide.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service