Skip to Content
Merck
All Photos(1)

Key Documents

05-5850

Sigma-Aldrich

Copper(II) chloride dihydrate

JIS special grade, ≥99.0%

Synonym(s):

Cupric chloride dihydrate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CuCl2 · 2H2O
CAS Number:
Molecular Weight:
170.48
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:

grade

JIS special grade

vapor density

>1 (vs air)

Assay

≥99.0%

form

solid

reaction suitability

reagent type: catalyst
core: copper

availability

available only in Japan

pH

3.0-3.8

mp

100 °C (dec.) (lit.)

SMILES string

Cl[Cu]Cl.[H]O[H].[H]O[H]

InChI

1S/2ClH.Cu.2H2O/h2*1H;;2*1H2/q;;+2;;/p-2

InChI key

MPTQRFCYZCXJFQ-UHFFFAOYSA-L

Looking for similar products? Visit Product Comparison Guide

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 2 - Eye Dam. 1 - Skin Irrit. 2

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

María S Islas et al.
Inorganic chemistry, 53(11), 5724-5737 (2014-05-16)
A new Cu(II) complex with the antihypertensive drug telmisartan, [Cu8Tlm16]·24H2O (CuTlm), was synthesized and characterized by elemental analysis and electronic, FTIR, Raman and electron paramagnetic resonance spectroscopy. The crystal structure (at 120 K) was solved by X-ray diffraction methods. The
Bin Wu et al.
Carbohydrate research, 351, 68-73 (2012-03-01)
Two new glucoside esters 1 and 2 were produced as stress metabolites in the fresh leaves of Portulaca oleracea, in response to abiotic stress elicitation by CuCl(2). A new sugar ester (3) and two known compounds (4 and 5) were
Iron(II)-catalyzed trifluoromethylation of potassium vinyltrifluoroborates.
Andrew T Parsons et al.
Angewandte Chemie (International ed. in English), 51(12), 2947-2950 (2012-02-14)
Francesca Cima et al.
Chemosphere, 89(1), 19-29 (2012-05-01)
After the widespread ban of TBT, due to its severe impact on coastal biocoenoses, mainly related to its immunosuppressive effects on both invertebrates and vertebrates, alternative biocides such as Cu(I) salts and the triazine Irgarol 1051, the latter previously used
Shuxia Zhou et al.
Pharmaceutical research, 30(5), 1311-1327 (2013-02-16)
Metal-catalyzed oxidation (MCO) of proteins is of primary concern in the development of biotherapeutics as it represents a prominent degradation pathway with potential undesired biological and biotherapeutic consequences. We developed a fluorogenic derivatization methodology to study the MCO of IgG1

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service