05-0900
Calcium oxide
SAJ first grade, ≥98.0%
Synonym(s):
Lime, Quicklime
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Linear Formula:
CaO
CAS Number:
Molecular Weight:
56.08
EC Number:
MDL number:
UNSPSC Code:
12352303
PubChem Substance ID:
Recommended Products
grade
SAJ first grade
Assay
≥98.0%
form
solid
reaction suitability
reagent type: catalyst
core: calcium
availability
available only in Japan
pH
12.6 (20)
bp
2850 °C (lit.)
density
3.3 g/mL at 25 °C (lit.)
SMILES string
O=[Ca]
InChI
1S/Ca.O
InChI key
ODINCKMPIJJUCX-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
Related Categories
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Eye Dam. 1 - Skin Irrit. 2 - STOT SE 3
Target Organs
Respiratory system
Storage Class Code
11 - Combustible Solids
WGK
WGK 1
Flash Point(F)
does not flash
Flash Point(C)
does not flash
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Arup Roy et al.
Journal of biomedical nanotechnology, 9(9), 1570-1578 (2013-08-29)
The high degree of microbial diseases and their multidrug resistant properties make the researchers to develop new class of antimicrobial agents. A modern and innovative approach of drug development is the use of metallic nanoparticles as new formulations of antimicrobial
Marco Faustini et al.
Nanoscale, 5(3), 984-990 (2012-12-19)
We report the simple preparation of ultra-thin self-assembled nanoperforated titanium calcium oxide films and their use as reactive nanomasks for selective dry etching of silicon. This novel reactive nanomask is composed of TiO(2) in which up to 50% of Ti
M Lopes et al.
Poultry science, 92(3), 638-644 (2013-02-26)
Testing different management practices can help to identify conditions that decrease or even eliminate pathogenic bacteria in poultry litter. A trial was conducted to evaluate the effects of daily manual stirring (rotation of the litter with a pitchfork) for the
E Belon et al.
The Science of the total environment, 439, 87-95 (2012-10-16)
The inputs of ten trace elements (As, Cd, Cu, Cr, Hg, Mo, Ni, Pb, Se, Zn) to French agricultural soils have been assessed. The six main sources considered were: pesticides, mineral fertilizers, animal manure, liming materials, sludge and composts and
N Husillos Rodríguez et al.
Waste management (New York, N.Y.), 32(3), 550-560 (2011-11-29)
This paper describes an industrial process for stabilising sewage sludge (SS) with lime and evaluates the viability of the stabilised product, denominated Neutral, as a raw material for the cement industry. Lime not only stabilised the sludge, raised the temperature
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service