Skip to Content
Merck
All Photos(1)

Key Documents

01-2790

Sigma-Aldrich

Aluminum nitrate nonahydrate

JIS special grade, ≥98.0%

Synonym(s):

Aluminium nitrate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Al(NO3)3 · 9H2O
CAS Number:
Molecular Weight:
375.13
EC Number:
MDL number:
UNSPSC Code:
12352311
PubChem Substance ID:

grade

JIS special grade

Assay

≥98.0%

form

solid

reaction suitability

reagent type: catalyst
core: aluminum

availability

available only in Japan

pH

2.5-3.5 (25 °C, 50 g/L)

mp

73 °C (lit.)

SMILES string

O.O.O.O.O.O.O.O.O.[Al+3].[O-][N+]([O-])=O.[O-][N+]([O-])=O.[O-][N+]([O-])=O

InChI

1S/Al.3NO3.9H2O/c;3*2-1(3)4;;;;;;;;;/h;;;;9*1H2/q+3;3*-1;;;;;;;;;

InChI key

SWCIQHXIXUMHKA-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Eye Dam. 1

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 1

Flash Point(F)

does not flash

Flash Point(C)

does not flash

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

L S Carneiro et al.
Dento maxillo facial radiology, 38(2), 73-78 (2009-01-30)
The aim of this study was to evaluate the performance of a quantitative method, based on pixel grey value measurements, for monitoring caries remineralization. Proximal radiographs of 11 patients (61 enamel caries lesions) were taken both before and after a
Jonathan H Seideman et al.
Assay and drug development technologies, 8(5), 602-614 (2010-07-28)
Alpha-particle-emitting elements are of increasing importance as environmental and occupational carcinogens, toxic components of radiation dispersal devices and accidents, and potent therapeutics in oncology. Alpha particle radiation differs from radiations of lower linear energy transfer in that it predominantly damages
L Trinkler et al.
Radiation protection dosimetry, 100(1-4), 313-316 (2002-10-18)
The ceramic material AlN-Y2O3 is proposed as a potential ultraviolet radiation (UVR) dosemeter using optically stimulated luminescence (OSL) and thermally stimulated luminescence (TL). Experimental studies have shown that AIN ceramics exhibit attractive characteristics suitable for practical UV dosimetry applications. The
Jason T Gatlin et al.
Inorganic chemistry, 47(4), 1267-1269 (2008-01-18)
Treatment of aluminum nitrate with an organic nitroso-containing compound yields the "flat", tridecameric nanocluster Al 13(mu 3-OH) 6(mu 2-OH) 18(H 2O) 24(NO 3) 15 ( Al 13 ) in good yield on a preparative scale under ambient conditions. Synthetic procedures
Qin Zheng Yang et al.
International journal of pharmaceutics, 326(1-2), 148-152 (2006-08-08)
Layered double hydroxides (LDHs) were investigated as cordycepin delivery nanocarriers for the first time in this study. Negatively charged biomolecule-cordycepin was intercalated in the gallery spaces of [Mg-Al-NO(3)] as the charge-compensating species, which was confirmed by the results of XRD

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service