Skip to Content
Merck
All Photos(5)

Key Documents

HPPCRPKRO

Roche

High Pure PCR Product Purification Kit

sufficient for 250 purifications (11732676001), sufficient for 50 purifications (11732668001), suitable for DNA purification

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41105500

usage

sufficient for 250 purifications (11732676001)
sufficient for 50 purifications (11732668001)

Quality Level

manufacturer/tradename

Roche

packaging

pkg of 250 purifications (11732676001)
pkg of 50 purifications (11732668001)

technique(s)

DNA purification: suitable

General description

Nucleic acids bind to the surface of the glass fiber fleece in the presence of a chaotropic salt (guanidine HCl). This allows the High Pure filter tube to specifically immobilize nucleic acids (both DNA and RNA) while they are freed of contaminants.

Capacity: The High Pure SpinFilter Tubes hold up to 700 μl sample volume.
Volume: 100 μl
Typical samples include:

  • Products from amplification or cDNA synthesis reactions
  • Enzymatically treated DNA
  • DNA from agarose slices
  • Dilute nucleic acid solutions
  • RNA from transcription reactions

The High Pure PCR Product Purification Kit efficiently and conveniently isolates PCR products from amplification reactions and purifies nucleic acids from other modification reactions. It is also recommended for the purification of cDNA.

Application

The High Pure PCR Product Purification Kit is designed for the preparation of concentrated, purified DNA, and can be used directly for most molecular biology applications:

  • Labeling
  • Sequencing
  • Cloning
  • Restriction enzyme digests
  • Alkaline phosphatase treatment
  • Kinase reactions


The kit eliminates primers, mineral oil, salts, unincorporated nucleotides, and thermostable DNA polymerases, which may inhibit subsequent enzymatic reactions. It can also be applied to concentrate dilute nucleic-acid solutions. Use one kit for a variety of applications.

The fast and simple High Pure protocols use a tabletop centrifuge to bind, wash, and elute the reaction product down to 10 μl (micro format) in as little as 10 minutes. The procedure conveniently eliminates a concentration step, and is ideal for downstream applications such as labeling, sequencing, cloning, ligation, or amplification using PCR.

Features and Benefits

  • Quickly purifies multiple PCR products
in <10 minutes, and purify DNA from agarose gel slices in <20 minutes.
  • Efficiently recover DNA fragments
>100 bp in length, and concentrate dilute nucleic acid solutions.
  • Minimize DNA loss
with a kit that removes contaminants without precipitation or other handling steps that degrade DNA.
  • Eliminate the use of hazardous organic compounds
such as cesium chloride, phenol, chloroform, and ethidium bromide

Components

  • Binding Buffer
  • Wash Buffer
  • Elution Buffer
  • High Pure Spin Filter Tubes (containing glass fiber fleece)
  • Collection Tubes

Quality

More than 70% recovery is obtained when 10 μg DNA Molecular Weight Marker VIII mixed with 16 μg bovine serum albumin are applied to the High Pure Filter Tubes. Gel electrophoresis of the DNA eluate confirms the complete removal of protein and DNA fragments smaller than 100 bp.

Preparation Note

The sample is mixed with a chaotropic salt and applied to the glass fiber fleece in a High Pure Spin Filter Tube. Under the buffer conditions used in the procedure, all nucleic acids (NA) in the sample bind to the glass fleece in the High Pure tube, while contaminating substances (salts, proteins, nucleotides, mineral oil and other contaminants) do not. Brief wash-and-spin steps readily remove these contaminants. Once purified, the NA can be easily eluted in a small volume of low salt buffer.

Other Notes

For life science research only. Not for use in diagnostic procedures.

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Chronic 3 - Eye Dam. 1 - Skin Corr. 1C

Supplementary Hazards

Storage Class Code

10 - Combustible liquids

WGK

WGK 2


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Gilda Eslami et al.
Iranian journal of parasitology, 14(3), 465-471 (2019-11-02)
The first line treatment for cutaneous leishmaniasis is pentavalent antimony such as sodium stibogluconate (pentostam) and meglumine antimonite (glucantime). One of the most important ways to uptake the drug is by a trans-membrane protein, called aquaglyceroporin encoded by Aquaglyceroprotein1 (LmAQP1).
The mitochondrial genomes of the acoelomorph worms Paratomella rubra, Isodiametra pulchra and Archaphanostoma ylvae
Robertson, et al.
Scientific Reports (2017)
Cancer-Testis Gene Biomarkers Discovered in Colon Cancer Patients
Almutairi, et al.
Genes, 13, 807-807 (22)

Protocols

Determine the labeling efficiency in terms of μg (expected yield of a standard labeling reaction is 20 μg of DIG labeled RNA per μg linearized template DNA after the DIG RNA labeling reaction).

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service