Skip to Content
Merck

CELLDETH-RO

Roche

Cell Death Detection ELISAPLUS

sufficient for 96 multiwell tests (11774425001), sufficient for 10 x 96 multiwell tests (11920685001)

Synonym(s):

ELISA

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

UNSPSC Code:
41116158
NACRES:
NA.54
Storage temp.:
2-8°C
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist

usage

sufficient for 10 x 96 multiwell tests (11920685001), sufficient for 96 multiwell tests (11774425001)

manufacturer/tradename

Roche

technique(s)

ELISA: suitable

detection method

photometric

storage temp.

2-8°C

General description

Use this kit for relative quantification of histone-complexed DNA fragments (mono- and oligonucleosomes) out of the cytoplasma of cells after the induction of apoptosis or when released from necrotic cells. Since the assay does not require prelabeling of cells, it can detect internucleosomal degradation of genomic DNA during apoptosis even in cells that do not proliferate in vitro, for example, freshly isolated tumor cells. The antibodies used in the assay are not species-specific, therefore, the kit may be used to assay cells from a wide variety of species.

Application

For research use only. Not for use in diagnostic procedures.

The Cell Death Detection ELISAPLUS photometric enzyme immunoassay is used for the quantitative in vitro determination of cytoplasmic histone-associated DNA fragments (mono- and oligonucleosomes) after induced cell death.
The kit contains a stop solution which allows users to terminate the substrate reaction and run the assay under defined conditions, making it suitable for use in high-throughput applications.

Biochem/physiol Actions

Anti-histone reacts with the histones H1, H2A, H2B, H3, and H4 of various species (e.g., human, mouse, rat, hamster, cow, opossum, Xenopus). Anti-DNA binds to single- and double-stranded DNA. Therefore, the ELISA allows the detection of mono- and oligonucleosomes from various species, and may be applied to measure apoptotic cell death in many different cell systems.

Features and Benefits

Assay time: 3 - 4 hours
Negative control: Depending on cell culture conditions, each exponentially growing permanent cell culture contains a certain amount of dead cells (typically approximately 3 - 8%). In the immunoassay, these inherent dead cells in the untreated sample (without a cell-death-inducing agent) will cause a certain absorbance value (negative control).
Positive control: A DNA-histone complex serves as a positive control.
Sample material: Cytoplasmic fractions (lysates) of cell lines, cells ex vivo, cell culture supernatants, and serum or plasma
  • One-step ELISA
  • High sensitivity: (5 x 102 cells/ml)
  • Fast performance: (3 - 4 hours)
  • Positive control included
  • No prelabeling of cells necessary
  • Nonradioactive assay system
  • No species restriction
  • Easy handling
  • Low background
  • Suppressed human anti-mouse factor
  • Function tested

Packaging

1 kit containing 10 components
  • 11774425001 (1 kit containing 10 components)
  • 11920685001 (1 kit containing 10 components)

Preparation Note

Working solution: Preparation of Working Solutions

Alongside the ready-to-use solutions supplied with this kit, you will need to prepare the following working solutions:
Note: Use only double-distilled water for reconstitution of lyophilizates.

Anti-Histone Biotin
Reconstitute the lyophilizate in 450 μl double-distilled water for 10 minutes and mix thoroughly.
Use: Component of the Immunoreagent.
Anti-DNA Peroxidase
Reconstitute the lyophilizate in 450 μl double-distilled water for 10 minutes and mix thoroughly.
Use: Component of the Immunoreagent.
Positive Control
Reconstitute the lyophilizate in 450 μl double-distilled water for 10 minutes and mix thoroughly.
Use: ELISA Step 1.
ABTS Tablets
Dependent on the number of samples tested, dissolve 1, 2, or 3 tablets from bottle 7 in 5, 10, or 15 ml Substrate Buffer.
Use: ELISA Step 5.
ABTS Stop Solution
If turbidity or a precipitate is visible, warm to 37 °C with shaking until the solution is clear.
Use: ELISA Step 6.
Storage conditions (working solution): Anti-Histone Biotin: 2 to 8 °C for 2 months
ABTS Tablets: 1 month stored protected from light. Allow to come to 15 to 25 °C before use.
ABTS Stop Solution: 2 to 8 °C until the expiration date printed on the label.
The exact detection limit of dying/dead cells in a particular sample strongly depends on the kinetics of cell death, the cytotoxic agent used, and the amount of affected cells in the total cell population. Using U937/camptothecin (CAM) as a cellular model system for cell death, the immunoassay allows the specific detection of mono- and oligonucleosomes in the cytoplasmic fraction of 125cell equivalents/well.

Other Notes

For life science research only. Not for use in diagnostic procedures.
Unit Conversion: 1 mU = 1 x 10-3 OD (1 mU = 0.001 OD).

Kit Components Only

Product No.
Description

  • Anti-histone-biotin antibody (clone H11-4)

  • Anti-DNA-POD antibody (clone MCA-33)

  • Positive Control

  • Incubation Buffer

  • Lysis Buffer

  • Substrate Buffer

  • ABTS Substrate Tablet

  • ABTS Stop Solution

  • Microplate (streptavidin-coated)

  • Adhesive Cover Foils

See All (10)

signalword

WarningDanger

Hazard Classifications

Aquatic Chronic 2 - Aquatic Chronic 3 - Eye Dam. 1 - Eye Irrit. 2 - Skin Sens. 1

Storage Class

8B - Non-combustible, corrosive hazardous materials

wgk

WGK 3

flash_point_c

does not flash


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Shuang Mei et al.
Endocrinology, 153(5), 2120-2129 (2012-03-01)
We addressed the link between excessive exposure to insulin and mitochondrion-derived oxidative stress in this study and found that prolonged exposure to insulin increased mitochondrial cholesterol in cultured hepatocytes and in mice and stimulated production of reactive oxygen species (ROS)
Bysani Chandrasekar et al.
The Journal of biological chemistry, 283(36), 24889-24898 (2008-07-18)
The adipocyte-derived cytokine adiponectin is known to exert anti-inflammatory and anti-apoptotic effects. In patients with atherosclerotic cardiovascular disease, circulating levels of adiponectin correlate inversely with those of the proinflammatory, proapoptotic cytokine interleukin (IL)-18. The opposing actions of IL-18 and adiponectin
Chu Zhang et al.
The Prostate, 71(2), 157-167 (2010-07-29)
Preferential bony metastasis of human prostate cancer (PCa) cells contributes to disease mortality and morbidity. Local factors in bone stromal extracellular matrix microenvironment affect tumor growth through paracrine interactions between tumor and stromal cells. Using co-culture and medium transfer, we

Global Trade Item Number

SKUGTIN
1177442500104061838261403
1192068500104061838261410

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service