Skip to Content
Merck
All Photos(1)

Key Documents

10786357001

Roche

Ribonuclease H (RNase H)

from Escherichia coli H 560 pol A1

Synonym(s):

rnase h

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352204

biological source

Escherichia coli ( H 560 pol A1)

Quality Level

Assay

100%

form

solution

specific activity

~40000 units/mg protein

packaging

pkg of 100 U

manufacturer/tradename

Roche

technique(s)

cDNA synthesis: suitable

color

colorless

optimum pH

7.5-9.1

solubility

water: miscible

suitability

suitable for molecular biology

NCBI accession no.

application(s)

life science and biopharma

foreign activity

RNase, none detected (up to 10 U with MS- II- RNA)
endonuclease ~10 units, none detected (using lambda-DNA)
nicking activity 10 units, none detected

shipped in

dry ice

storage temp.

−20°C (−15°C to −25°C)

Gene Information

Escherichia coli ... rnhA(946955)

General description

Nonspecific endoribonuclease that specifically cleaves RNA in RNA:DNA hybrids. A minimum of four continuous base pairs (RNA:DNA) is required for activity. RNase H cleaves RNA to release 5′-oligoribonucleotides.

Source: E. coli H560 pol A1
Storage Buffer: 25 mM Tris-HCl, 50 mM KCl, 1 mM dithiothreitol, 0.1 mM EDTA, 50% glycerol (v/v), pH 8.0 (+4°C)
Volume Activity: 1 x 103 U/ml assayed according to Hillenbrand & Staudenbauer.
Ribonuclease H (RNase H) is a nonspecific endoribonuclease, localized to the nucleus and cytoplasm. It is ubiquitously found and widely present among many organisms including viruses and human.

Application

Ribonuclease H (RNase H) has been used for:
  • In vivo RNA-primed initiation of DNA synthesis
  • Elimination of mRNA during second-strand cDNA synthesis
  • Site-specific cleavage of RNA
  • Detection of RNA:DNA regions in double-stranded DNA of natural origin
  • Removal of poly (A) sequences of mRNA if oligo (dT) is present
  • RNA extraction and quantitative reverse transcriptase polymerase chain reaction (RT-PCR)

Biochem/physiol Actions

Ribonuclease H (RNase H) specifically cleaves RNA in RNA:DNA hybrids. A minimum of four continuous base pairs (RNA:DNA) is required for activity. RNase H cleaves RNA to release 5′-oligoribonucleotides. RNase H is associated with nucleic acid immunity. Using RNase H for degrading mRNA results in 80% depletion of mRNA and protein expression. RNase H recognizes the start codon and the 3′ and 5′ untranslated regions. This enzyme participates in DNA replication.

Features and Benefits

  • Eliminate potential sources of PCR errors.
  • Increase accessibility of primers during subsequent PCR.

Quality

Absence of endonucleases, nicking activities, and ribonucleases.

Unit Definition

RNase H is assayed according to Hillenbrand and Staudenbauer. One unit of RNase H is the amount of enzyme which produces 1 nmol acid-soluble ribonucleotides from[3H] poly(A) x poly(dT) in 20 minutes at +37 °C under the stated assay conditions.

Volume Activity: Approximately 1 U/μl

Preparation Note

Activator: The enzyme has its maximal activity in presence of SH-reagents

Storage and Stability

Store at -15–-25 °C. (unopened)

Other Notes

For life science research only. Not for use in diagnostic procedures. Using RNase H after the cDNA synthesis step can increase the sensitivity of a two-step RT-PCR assay.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

does not flash

Flash Point(C)

does not flash


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Nucleic acid immunity
Hartmann G
Advances in Immunology null
Amandine Bonnet et al.
Molecular cell, 67(4), 608-621 (2017-08-02)
Transcription is a source of genetic instability that can notably result from the formation of genotoxic DNA:RNA hybrids, or R-loops, between the nascent mRNA and its template. Here we report an unexpected function for introns in counteracting R-loop accumulation in
Antisense oligonucleotides and rna interference
Kher G, et al.
Chalcogenide Lett null
β-catenin activity in late hypertrophic chondrocytes locally orchestrates osteoblastogenesis and osteoclastogenesis.
Houben A, et al.
Development, dev-137489 (2016)
Antisense therapy
Crooke ST
The Cytokine Handbook null

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service