Skip to Content
Merck
All Photos(1)

Key Documents

04716728001

Roche

DNase I recombinant, RNase-free

from bovine pancreas, expressed in Pichia pastoris

Sign Into View Organizational & Contract Pricing


About This Item

Enzyme Commission number:
UNSPSC Code:
12352204
NACRES:
NA.21
form:
solution
solubility:
water: soluble

biological source

bovine pancreas

Quality Level

recombinant

expressed in Pichia pastoris

form

solution

mol wt

~39 kDa

packaging

pkg of 10,000 U

manufacturer/tradename

Roche

optimum pH

7.0-8.0

solubility

water: soluble

General description

Recombinant DNase I is a DNA-specific endonuclease.The enzyme catalyzes the degradation of both double- and single-stranded DNA randomly by hydrolyzing phosphodiester linkages to DNA, resulting in a mixture of oligo- and mononucleotides. All material used during the production process of DNase I recombinant is non-animal sourced, resulting in an animal-free product.

Contents
  • Recombinant DNase I, RNase-free, 10 U/μl
  • Incubation Buffer, 10x concentrated

Specificity

Heat inactivation: One unit DNase I recombinant, RNase-free is heat-inactivated by 10 minutes incubation at 75 °C.

Important Note: Alternatively, DNase I recombinant, RNase-free can be inactivated and removed by phenol extraction according to standard protocols, e.g., Current Protocols in Molecular Biology.

Application

DNase I recombinant, RNase-free may be used to degrade DNA in applications that are sensitive to the presence of RNase. For example, DNase I is frequently used to:
  • Remove genomic DNA from RNA preparations prior to RT-PCR
  • Isolate DNA-free RNA after in vitro transcription reactions
  • Perform nick translations
  • Map DNase-sensitive regions in eukaryotic DNA

Features and Benefits

  • Eliminates DNA contamination from any RNA sample
  • Contains no detectable RNase or protease activity
  • Can be heat inactivated, thereby eliminating the need for organic extraction
  • Shipped with an optimized incubation buffer, which supports maximum DNase activity
  • Produced via an entirely animal-free process, to eliminate any risks associated with animal-derived material

Packaging

1 kit containing 2 components

Quality

Absence of contaminants: Each lot is tested to ensure the absence of RNases and proteases according to the current Quality Control procedures.

Specifications

Glycosylated form
Recombinant DNase I is heterogeneously N-glycosylated, so it appears as two bands in gel electrophoresis.
Divalent ion requirement
DNase I requires divalent cations for maximum activity. The DNA-specific endonuclease is activated by ions such as magnesium ions and is stimulated by calcium ions. Therefore, the enzyme is inhibited by metal chelating agents like EDTA.

Unit Definition

One unit is the enzyme activity that effects an absorbance increase of 0.001/minute under assay conditions in 1 ml at 260 nm.
Assay conditions:
Volume activity is determined according to the following assay mixture. 100 μg calf thymus DNA is incubated in 2.5 ml 1x incubation buffer with 40 to 70 units DNase I recombinant, RNase-free at +25 °C. The absorbance increase is measured at 260 nm.

Preparation Note

Activator: Bivalent metal ions
Working solution: Storage Buffer: 20 mM Tris-HCl, 50 mM NaCl, 2 mM CaCl2, 2 mM MgCl2, 1 mM dithioerythritol, 0.1 mg/ml Pefabloc SC, 50% glycerol (v/v), pH 7.6 (at 4 °C).
Incubation Buffer (10x): 400 mM Tris-HCl, 100 mM NaCl, 60 mM MgCl2, 10 mM CaCl2, pH 7.9.
Enzyme Dilution Buffer: 25 mM Tris-HCl, 50% glycerol (v/v), pH 7.6 (at 4 °C).

Storage and Stability

Store undiluted enzyme solution at -15 to -25°C; storage buffer at 4 °C.

Other Notes

For life science research only. Not for use in diagnostic procedures.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

does not flash

Flash Point(C)

does not flash


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Christina N Cheng et al.
Journal of visualized experiments : JoVE, (89)(89), doi:10-doi:10 (2014-08-01)
The zebrafish embryo is now commonly used for basic and biomedical research to investigate the genetic control of developmental processes and to model congenital abnormalities. During the first day of life, the zebrafish embryo progresses through many developmental stages including
In Kyoung Mah et al.
Developmental biology, 416(1), 82-97 (2016-06-18)
Polarized epithelia define boundaries, spaces, and cavities within organisms. Cavitation, a process by which multicellular hollow balls or tubes are produced, is typically associated with the formation of organized epithelia. In order for these epithelial layers to form, cells must
Lucas R Smith et al.
Cellular physiology and biochemistry : international journal of experimental cellular physiology, biochemistry, and pharmacology, 54(3), 333-353 (2020-04-11)
Cell migration and extracellular matrix remodeling underlie normal mammalian development and growth as well as pathologic tumor invasion. Skeletal muscle is no exception, where satellite cell migration replenishes nuclear content in damaged tissue and extracellular matrix reforms during regeneration. A
Kirsten Richter et al.
The Journal of biological chemistry, 295(23), 7849-7864 (2020-04-23)
Activation of the T cell receptor (TCR) results in binding of the adapter protein Nck (noncatalytic region of tyrosine kinase) to the CD3ϵ subunit of the TCR. The interaction was suggested to be important for the amplification of TCR signals
Kwok-Fong Chan et al.
Molecules (Basel, Switzerland), 25(24) (2020-12-17)
The ongoing development of drug resistance in HIV continues to push for the need of alternative drug targets in inhibiting HIV. One such target is the Reverse transcriptase (RT) enzyme which is unique and critical in the viral life cycle-a

Articles

Organoid culture products to generate tissue and stem cell derived 3D brain, intestinal, gut, lung and cancer tumor organoid models.

Organoid culture products to generate tissue and stem cell derived 3D brain, intestinal, gut, lung and cancer tumor organoid models.

Organoid culture products to generate tissue and stem cell derived 3D brain, intestinal, gut, lung and cancer tumor organoid models.

Organoid culture products to generate tissue and stem cell derived 3D brain, intestinal, gut, lung and cancer tumor organoid models.

Protocols

DNase I from bovine pancreas is a glycoprotein of Mr 37000. A special procedure is used to remove RNases from the DNase preparation.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service