VPMKVALNB9
Viresolve® Pro Device
Parvovirus removal filter, Micro 40 kit, filtration area 3.4 cm²
Synonym(s):
Viresolve® Pro Device Micro 40 Kit
About This Item
material
polyethersulfone membrane ((PES))
white PVDF device (molded parts)
Quality Level
parameter
4.1 bar max. pressure (60 psid) at 25 °C (Samples were integral based on an Air/ Water Diffusion Test, before and after a forward stress.)
H × diam. × filtration area
4.03 cm (1.59 in.) × 4.37 cm (1.72 in.) × 3.4 cm2
filtration area
3.4 cm2
weight
14.73 gempty
impurities
may contain <0.25 EU/mL bacterial endotoxins (LAL test)
fitting
female Luer-Lok® inlet (inlet and vent)
male Luer outlet (Slip)
storage temp.
no temp limit
Looking for similar products? Visit Product Comparison Guide
General description
*Viresolve® Pro Devices are built with two membrane layers. Each Viresolve® Pro Device Micro 40 is packaged in a blister pack and labeled with the device lot and the top layer membrane lot/roll number.
Application
Features and Benefits
- Small scale devices for process development, optimization and validation studies
- Membrane based adsorptive prefilters to remove protein aggregates and other fouling species
- Nine units per kit (one Membrane lot)
Other Notes
Legal Information
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service