Skip to Content
Merck
All Photos(3)

Key Documents

UFC205024

Millipore

Amicon® Ultra Centrifugal Filter, 50 kDa MWCO

greener alternative

sample volume 2 mL, regenerated cellulose membrane, MWCO 50 kDa

Synonym(s):

Centrifugal concentrator, 50 kDa MWCO, 2 mL sample volume

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41104916
eCl@ss:
36100101
NACRES:
NB.24

material

polypropylene tube (for collection)
regenerated cellulose membrane
styrene-butadiene housing

Quality Level

sterility

non-sterile

product line

Amicon®

manufacturer/tradename

Amicon® Ultra

sustainability

Greener Alternative Product

parameter

2 mL sample volume

technique(s)

protein extraction: suitable
protein purification: suitable

L

119.7 mm

diam.

15.9 mm

filtration area

1 cm2

working volume

2 mL

pore size

50 kDa MWCO

greener alternative category

shipped in

ambient

General description

Amicon® Ultra-2 mL centrifugal devices with Ultracel® 50,000 Dalton molecular weight cutoff filters provide fast ultrafiltration, with the capability for high concentration factors and easy concentrate recovery from dilute and complex sample matrices. For example, BSA (67 kDa) at 1 mg/ml initial concentration is typically retained at 97% .
The vertical design and available membrane surface area provide fast sample processing, high sample recovery (typically greater than 90% of dilute starting solution), and the capability for 50-fold concentration. Solute polarization and subsequent fouling of the membrane are minimized by the vertical design, and a physical deadstop in the filter device prevents spinning to dryness and potential sample loss. Efficient recovery of the concentrated sample (retained species) is achieved by a convenient reverse spin step after collecting the filtrate.
The device can be spun in a swinging bucket or fixed angle rotor. Amicon® Ultra-2 mL devices are supplied non-sterile and are for single use only. With their vertical membrane design and reverse spin, Amicon® Ultra 2 mL filters deliver the highest recovery with shortest spin times.
We are committed to bringing you Greener Alternative Products, which have been re-engineered to enable more sustainable research solutions. The inner filtration device is made from bio-circular material allocated using the ISCC mass balance approach. Click here for more information.

Application

  • Ultrafiltration with 50,000 Dalton molecular weight cutoff Ultracel® membrane.
  • Concentration of biological samples containing antigens, antibodies, enzymes, nucleic acids, or microorganisms.
  • Purification of macromolecular components found in tissue culture extracts or cell lysates.
  • Primer removal, PCR cleanup, removal of linkers or macromolecular labels from a reaction mix, and protein removal prior to HPLC.
  • Desalting, buffer exchange and protein dialysis.
  • Size exclusion and protein fractionation.

Features and Benefits

  • Ultracel® regenerated cellulose membrane with 50 kDa NMWCO delivers retentate recovery of greater than 90%
  • Concentrates 2 mL down to 15-70 μL
  • 98% sample recovery
  • Fast processing time: 10 to 30-minute spin times
  • Engineered dead stop
  • Reverse spin capability provides consistent recoveries

Linkage

Replaces: 4224

Physical form

Color Code: Clear

Legal Information

Amicon is a registered trademark of Merck KGaA, Darmstadt, Germany
ULTRACEL is a registered trademark of Merck KGaA, Darmstadt, Germany

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

Explore ultrafiltration membrane principles and benefits for protein sample preparation, including solvent exchange and sample concentration.

Explore ultrafiltration membrane principles and benefits for protein sample preparation, including solvent exchange and sample concentration.

Explore ultrafiltration membrane principles and benefits for protein sample preparation, including solvent exchange and sample concentration.

Explore ultrafiltration membrane principles and benefits for protein sample preparation, including solvent exchange and sample concentration.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service