TX1202
Triethylamine
HPLC
Synonym(s):
Triethylamine, acid
About This Item
Recommended Products
vapor pressure
72 hPa ( 20 °C)
Quality Level
Assay
≥99.5% (GC)
form
liquid
technique(s)
HPLC: suitable
color
clearAPHA: ≤20
pH
12.7 (15 °C, 100 g/L in H2O)
bp
90 °C/1013 hPa
mp
-115 °C
transition temp
flash point -11 °C
solubility
133 g/L
density
0.73 g/cm3 at 20 °C
UV absorption
λ: 254 nm Amax: ≤0.10
λ: 280 nm Amax: ≤0.04
λ: 300 nm Amax: ≤0.02
shipped in
ambient
storage temp.
room temp
SMILES string
N(CC)(CC)CC
InChI
1S/C6H15N/c1-4-7(5-2)6-3/h4-6H2,1-3H3
InChI key
ZMANZCXQSJIPKH-UHFFFAOYSA-N
Application
Analysis Note
Assay: 99.5% min
Color (APHA): 20 max
Identity (IR-spectrum): Conforms
UV Absorbance (0.1%, water, 254 nm): 0.10AU max
UV Absorbance (0.1%, water, 280 nm): 0.04AU max
UV Absorbance (0.1%, water, 300 nm): 0.02AU max
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 4 Oral - Eye Dam. 1 - Flam. Liq. 2 - Skin Corr. 1A - STOT SE 3
Target Organs
Respiratory system
Storage Class Code
3 - Flammable liquids
WGK
WGK 1
Flash Point(F)
12.2 °F - closed cup
Flash Point(C)
-11 °C - closed cup
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service