Skip to Content
Merck
All Photos(4)

Key Documents

SCC131

Sigma-Aldrich

A2EN Human Endocervical Epithelial Cell Line

Human

Synonym(s):

A2EN Cell Line

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106514
eCl@ss:
32011203

product name

A2EN Human Endocervical Epithelial Cell Line, A2EN endocervical epithelial cell line can be used to study sexually transmitted pathogen infections, pathogenesis and the innate immune response.

biological source

human

Quality Level

technique(s)

cell culture | mammalian: suitable

General description

A2EN is an immortalized human epithelial cell line derived from human primary epithelial cells expanded from an explant of human endocervical tissue. The primary endocervical epithelial cells were immortalized with lentiviral vectors carrying the HPV E6/E7 genes. A2EN cells can generate a transepithelial electrical resistance (TEER) when grown on cell culture inserts and generate differential apical and basolateral cytokine profiles upon stimulation with various toll-like receptor (TLR) agonists.
The endocervical epithelium is a single cell layer lining the endocervical canal that separates the lower female reproductive tract from the uterus and Fallopian tubes. Endocervical epithelial cells secrete mucus and innate immune mediators that facilitate reproduction as well as protect from pathogen invasion. The endocervix is a major site of transmission for multiple sexually transmitted organisms, such as Chlamydia trachomatis, Neisseria gonorrhoeae, Mycoplasma genitalium and human immunodeficiency virus (HIV).

Cell Line Description

Epithelial Cells

Application

A2EN endocervical epithelial cell line can be used to study sexually transmitted pathogen infections, pathogenesis and the innate immune response.
Research Category
Infectious Diseases

Immune Response
This product is intended for sale and sold solely for internal non-commercial research use per the terms of the “Restricted Use Agreement” as detailed in the product documentation. For information regarding any other uses, please contact licensing@emdmillipore.com.

Quality

• Each vial contains ≥ 1X106 viable cells.
• Cells are tested negative for Epstein-Barr virus, HPV-16, HPV-18, Hepatitis A, C, Herpesvirus type 6, 7, 8 and HIV-1 & 2 viruses by PCR
• Cells are negative for mycoplasma contamination.
• Each lot of cells is genotyped by STR analysis to verify the unique identity of the cell line.

Storage and Stability

Store in liquid nitrogen. The cells can be cultured for at least 10 passages after initial thawing without significantly affecting the cell marker expression and functionality.

Disclaimer

This product contains genetically modified organisms (GMO). Within the EU GMOs are regulated by Directives 2001/18/EC and 2009/41/EC of the European Parliament and of the Council and their national implementation in the member States respectively. This legislation obliges {HCompany} to request certain information about you and the establishment where the GMOs are being handled. Click here for Enduser Declaration (EUD) Form.

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

does not flash

Flash Point(C)

does not flash


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Melissa M Herbst-Kralovetz et al.
American journal of reproductive immunology (New York, N.Y. : 1989), 59(3), 212-224 (2008-01-19)
To better understand innate immune responses to sexually-transmitted infection (STI) and the appropriateness of epithelial cell (EC) models of the vaginal and cervical mucosa, quantified toll-like receptor (TLR) expression from a population of women is needed. TLR gene expression was
Lyndsey R Buckner et al.
Journal of reproductive immunology, 92(1-2), 8-20 (2011-09-29)
The endocervix in the female reproductive tract (FRT) is susceptible to sexually transmitted pathogens such as Chlamydia trachomatis and Neisseria gonorrhoeae. Endocervical epithelial cells in vivo make innate immune mediators that likely aid in the protection from these pathogens. In
Jaganathan Kowshik et al.
IUBMB life, 71(10), 1595-1610 (2019-06-30)
Aberrant activation of the PI3K/Akt signalling pathway, a major driving force of diverse cellular processes has been implicated in tumour development and progression. Here, we report that astaxanthin (AXT), a potent antioxidant ketocarotenoid prevents cancer hallmarks by inhibiting PI3K/Akt and
Mohammad Iqbal Rather et al.
The Journal of biological chemistry, 289(2), 968-976 (2013-11-22)
The Wilms tumor 1 gene (WT1) can either repress or induce the expression of genes. Inconsistent with its tumor suppressor role, elevated WT1 levels have been observed in leukemia and solid tumors. WT1 has also been suggested to act as

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service