Skip to Content
Merck
All Photos(1)

Key Documents

SCC069

Sigma-Aldrich

HSC-T6 Rat Hepatic Stellate Cell Line

Rat

Synonym(s):

HSC, Perisinusoidal cells, Ito cells, Hepatic lipocytes, Hepatic pericytes

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106514
eCl@ss:
32011203
NACRES:
NA.81

product name

HSC-T6 Rat Hepatic Stellate Cell Line, The HSC-T6 rat hepatic stellate cell line is a valuable cell model for studies of retinoid metabolism based on their similar retinoid phenotype as primary cells.

biological source

rat

Quality Level

technique(s)

cell based assay: suitable
cell culture | mammalian: suitable

shipped in

ambient

General description

Hepatocytes and hepatic stellate cells play important roles in retinoid uptake, storage, mobilization and metabolism. Hepatic stellate cells represent only 5-7% of the total cells present in the liver. Isolation of primary stellate cells is thus time-consuming and often accompanied by low yields and considerable lot-to-lot variations. HSC-T6 Rat Hepatic Stellate Cell Line is an immortalized line isolated from SV40 large T-antigen transformed primary stellate cells of male Sprague-Dawley retired breeder rats. HSC-T6 Rat Hepatic Stellate Cell line is a useful model for the study of hepatic retinoid storage and metabolism.

Cell Line Description

Hepatocyte Cells

Application

Research Category
Metabolism

Infectious Diseases

Inflammation & Immunology

Stem Cell Research
Research Sub Category
Liver Physiology
This product is intended for sale and sold solely to academic institutions for internal academic research use per the terms of the “Academic Use Agreement” as detailed in the product documentation. For information regarding any other use, please contact licensing@emdmillipore.com.

Components

1) ≥1X106 viable HSC-T6 Rat Hepatic Stellate Cells: (Catalog No. SCC069). Store in liquid nitrogen.

Quality

• Each vial contains ≥ 1X106 viable cells.
• Cells tested negative for infectious diseases by a Mouse/Rat Comprehensive CLEAR panel PCR panel by Charles River Animal Diagnostic Services.
• Cells tested negative for mycoplasma contamination.

Storage and Stability

HSC-T6 Rat Hepatic Stellate Cells should be stored in liquid nitrogen. The cells can be cultured for at least 10 passages after initial thawing without significantly affecting the cell marker expression and functionality.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ryuichi Noguchi et al.
Molecular medicine reports, 22(4), 2948-2956 (2020-09-19)
An orally bioavailable small molecule inhibitor of plasminogen activator inhibitor‑1 (PAI‑1) is currently being clinically assessed as a novel antithrombotic agent. Although PAI‑1 is known to serve a key role in the pathogenesis of metabolic syndrome (MetS) including nonalcoholic steatohepatitis
Scott L Friedman et al.
Methods in molecular biology (Clifton, N.J.), 2669, 129-162 (2023-05-29)
Hepatic stellate cells (HSCs) are the major cellular source of extracellular matrix production in the liver. Therefore, this cell population has received considerable attention in studies investigating fundamental features of hepatic fibrosis. However, the limited supply and ever-increasing demand for
Jiunn-Sheng Wu et al.
Evidence-based complementary and alternative medicine : eCAM, 2020, 8426051-8426051 (2020-07-28)
Hepatic stellate cell (HSC) activation is a vital driver of liver fibrosis. Recent research efforts have emphasized the clearance of activated HSCs by apoptosis, senescence, or reversion to the quiescent state. LPS induces human HSC activation directly and contributes to
Hongxia Liang et al.
ACS nano, 12(7), 6536-6544 (2018-06-27)
Acute liver failure is a critical condition characterized by global hepatocyte death and often time needs a liver transplantation. Such treatment is largely limited by donor organ shortage. Stem cell therapy offers a promising option to patients with acute liver
Hsiao-Hsuan Wang et al.
EBioMedicine, 64, 103213-103213 (2021-01-29)
Interleukin-24 (IL-24) binds to two kinds of receptor complexes, namely IL-20R1/IL-20R2 and IL-20R2/IL-22R1, which are also bound by IL-20. IL-20 plays a detrimental role in liver fibrosis. Due to the sharing of receptor complexes, we aimed to determine whether IL-24

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service