Skip to Content
Merck
All Photos(1)

Key Documents

PXC100C50

Millipore

Pellicon® XL Cassette with Ultracel® Membrane

filtration area 50 cm2, NMWCO 100 kDA

Synonym(s):

0.005 m2, 100 kDa, Pellicon XL, Pellicon® XL Cassette with Ultracel® Membrane, Ultracel, Ultrafiltration Module

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41104903

material

composite regenerated cellulose (CRC) membrane
polypropylene housing
polypropylene screen ((feed))
polypropylene screen ((permeate))

Quality Level

product line

Pellicon®

technique(s)

protein purification: suitable
tangential flow filtration (TFF): suitable (analytical sample preparation)

L × W

18.8 cm (7.4 in.) × 3 cm (1.2 in.)

filtration area

50 cm2

matrix

Ultracel®

pore size

100 kDa pore size

pH

2-12

Looking for similar products? Visit Product Comparison Guide

General description

  • Device Configuration: Cassette
  • Filter Code: PLCHK
  • Filter Type: Ultrafiltration

Features and Benefits

Type C screen (coarse screen)

Packaging

Wet, preserved

Other Notes

These products are manufactured in a facility which adheres to FDA Good Manufacturing Practices.

Legal Information

Pellicon is a registered trademark of Merck KGaA, Darmstadt, Germany
ULTRACEL is a registered trademark of Merck KGaA, Darmstadt, Germany

Not finding the right product?  

Try our Product Selector Tool.

Pictograms

Corrosion

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Met. Corr. 1

Storage Class Code

8B - Non-combustible corrosive hazardous materials

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service