Skip to Content
Merck
All Photos(1)

Key Documents

PLTK15005

Millipore

Regenerated Cellulose Membrane Filter, 30 kDa NMWCO

Ultracel®, filter diam. 150 mm

Synonym(s):

Ultrafiltration Discs, 30 kDa NMW

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41104903
eCl@ss:
32031602
NACRES:
JA.14
pore size:
30 kDa NMWCO
material:
regenerated cellulose membrane
filter diam.:
150 mm
filtration area:
177 cm2

material

regenerated cellulose membrane

Quality Level

manufacturer/tradename

Ultracel®

technique(s)

protein clean-up: suitable (desalting)
protein purification: suitable (concentration)

filter diam.

150 mm

filtration area

177 cm2

matrix

Ultracel®

pore size

30 kDa NMWCO

shipped in

ambient

Application

Ultrafiltration Discs, 30 kDa NMW has been used:
  • in cell-material interaction studies
  • as a modified fouling index-ultrafiltration (MFI-UF) test membrane to study the effect of silica concentration on the MFI-UF
  • in the feed fouling monitor (FFM) experiments to capture the model foulants

Legal Information

ULTRACEL is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ming Ni et al.
Biomaterials, 33(2), 353-364 (2011-10-25)
We developed a library of industrial materials, which can be applied to any adherent cell type for determining cell-material interactions. Bulk and surface chemistry as well as other material properties were characterized. The library covered broad ranges of various material
The fouling potential of colloidal silica and humic acid and their mixtures
Taheri AH, et al.
Journal of Membrane Science, 433, 112-120 (2013)
Prediction of reverse osmosis fouling using the feed fouling monitor and salt tracer response technique
Taheri AH and Sim, LN and Chong, TH and Krantz, WB and Fane, AG
Journal of Membrane Science, 475, 433-444 (2015)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service