Skip to Content
Merck
All Photos(1)

Key Documents

PLBC04710

Millipore

Regenerated Cellulose Membrane Filter, 3 kDa NMWCO

Ultracel®, filter diam. 47 mm

Synonym(s):

Ultrafiltration Discs, 3 kDa NMW, Ultrafiltration membrane discs

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41104903
eCl@ss:
32031602
NACRES:
NB.24

material

regenerated cellulose membrane

Quality Level

manufacturer/tradename

Ultracel®

technique(s)

protein clean-up: suitable (desalting)
protein purification: suitable (concentration)

diam.

47 mm

filter diam.

47 mm

filtration area

17.3 cm2

matrix

Ultracel®

pore size

3 kDa NMWCO

shipped in

ambient

Looking for similar products? Visit Product Comparison Guide

Legal Information

ULTRACEL is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service