Skip to Content
Merck
All Photos(2)

Key Documents

MRCPRT010

Millipore

Microcon® Centrifugal Filters

NMWCO 10 kDa, Ultracel® regenerated cellulose membrane (low binding), sample volume 0.5 mL

Synonym(s):

Centrifuge concentrator

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41104916
eCl@ss:
36100101
NACRES:
NB.24

material

Ultracel® regenerated cellulose membrane (low binding)
polyacetal base (membrane support)
polycarbonate device top
polypropylene tube (for filtrate/concentrate)
silicone O-ring (medical-grade)

Quality Level

sterility

non-sterile

feature

holdup volume> 10 μL

packaging

pkg of 100 ea

manufacturer/tradename

Microcon®

parameter

0.5 mL sample volume

technique(s)

DNA purification: suitable
RNA purification: suitable
protein extraction: suitable
protein purification: suitable

L

4.50 cm (1.8 in.)

diam.

12.3 mm

filtration area

0.32 cm2

working volume

0.5 mL

pore size

10 kDa NMWCO

shipped in

ambient

General description

Filter Code: PL, Ultracel® low binding regenerated cellulose
Filter Type: Ultrafiltration
Simply and efficiently concentrate and desalt solutions of any macromolecule, using any centrifuge that can accept 1.5 mL tubes. With the low-binding Ultracel membrane, Microcon filters offer:
•Typical recoveries of greater than 95%, even for dilute solutions
•Reverse spin to maximize recovery, even in the smallest samples
•Convenient storage of filtrate or concentrated sample in standard microfuge tube
•Concentration factors up to 100X

Features:
•High recovery for small volumes with reverse spin (concentration factor less than 20X)
•Low-binding Ultracel membrane
•Fast processing

Application

This 0.5 mL centrifugal filter enables the concentration of biomolecules with a membrane NMWL of 10 kDa. Micron centrifugal filters are ideal for DNA & protein concentration & desalting.

Features and Benefits

Minimum Final Concentrate Volume: 50 µL

Linkage

Replaces: 42407

Physical form

Color Code: Green

Legal Information

MICROCON is a registered trademark of Merck KGaA, Darmstadt, Germany
ULTRACEL is a registered trademark of Merck KGaA, Darmstadt, Germany

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Lilly Toker et al.
Chemico-biological interactions, 319, 109007-109007 (2020-02-23)
Acetylcholinesterase (AChE) terminates cholinergic neurotransmission by hydrolyzing acetylcholine. The collagen-tailed AChE tetramer is a product of 2 genes, ACHE and ColQ. The AChE tetramer consists of 4 identical AChE subunits and one polyproline-rich peptide, whose function is to hold the
Ruth Fair-Mäkelä et al.
Mucosal immunology, 13(2), 245-256 (2019-11-28)
All leukocytes can get entrance into the draining lymph nodes via the afferent lymphatics but only lymphoid cells can leave the nodes. The molecular mechanisms behind this phenomenon have remained unknown. We employed genome wide microarray analyses of the subcapsular
Anthony O Beas et al.
Nature communications, 11(1), 2790-2790 (2020-06-05)
Age-dependent changes in metabolism can manifest as cellular lipid accumulation, but how this accumulation is regulated or impacts longevity is poorly understood. We find that Saccharomyces cerevisiae accumulate lipid droplets (LDs) during aging. We also find that over-expressing BNA2, the
Yaowei Liu et al.
Food chemistry, 314, 126177-126177 (2020-01-20)
This study mainly investigated changes in milk serum proteins by simulating the industrial processing of whole milk powder using a label-free proteomic approach. A total of 391 proteins were identified, 89 of which were quantified in all groups. Compared with
Gulzhan Trimova et al.
Arthritis research & therapy, 22(1), 24-24 (2020-02-14)
14-3-3η is an intracellular protein also detected in the serum and synovial fluid of patients with rheumatoid arthritis (RA). It is closely related to disease activity and anti-cyclic citrullinated peptide antibody levels. However, the main source of 14-3-3η and the

Related Content

Microcon® Centrifugal Filters simply and efficiently concentrate and desalt solutions of DNA, RNA, protein or other macromolecules, using any centrifuge that can accept 1.5 mL tubes.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service