Skip to Content
Merck
All Photos(2)

Key Documents

MMMP3MAG-79K

Millipore

MILLIPLEX® Mouse MMP Magnetic Bead Panel 3 - Immunology Multiplex Assay

MMP Bead-Based Multiplex Assays using the Luminex technology enables the simultaneous analysis of multiple MMP biomarkers in mouse cell culture samples.

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12161503
eCl@ss:
32161000
NACRES:
NA.84

Quality Level

species reactivity

mouse

manufacturer/tradename

Milliplex®

assay range

standard curve range: 12-50,000 pg/mL
(MMP-8)

standard curve range: 12-50,000 pg/mL
(proMMP-9)

standard curve range: 2-10,000 pg/mL
(MMP-3)

standard curve range: 49-200,000 pg/mL
(MMP-2)

standard curve range: 5-20,000 pg/mL
(MMP-12)

technique(s)

multiplexing: suitable

detection method

fluorometric (Luminex xMAP)

shipped in

wet ice

General description

The MMPs (matrix metalloproteinases), a family of zinc proteases responsible for the breakdown of extracellular matrix (ECM), play a key role in normal physiological processes, such as embryonic development and tissue morphogenesis, tissue and bone remodeling, wound healing, and angiogenesis. These processes rely on MMPs′ role in the cleavage of cell surface receptors, the release of apoptotic ligands, cell proliferation and differentiation, and chemokine activity modulation. Similar in structure, MMPs are synthesized and secreted as inactive pro-enzymes that require proteolytic cleavage for activation. This process can be mediated by serine proteases or other MMPs. An increase in MMP expression occurs in response to a wide range of stimuli, including adhesion molecules, growth factors, cytokines, and hormones. Regulation of MMP activity is controlled primarily by TIMPs (tissue inhibitors of metalloproteinases). Therefore, disruption of the MMP/TIMP balance can result in autoimmune disease, arthritis, cardiovascular disease, and tumor growth and metastasis.

MILLIPLEX® Mouse MMP Bead Panel 3, MMMP3MAG-79K, is a 5 plex kit to be used for the simultaneous quantification of any or all of the following analytes in tissue culture samples: MMP-2, MMP-3, MMP-8, proMMP-9, and MMP-12.

Panel Type: Cytokines/Chemokines

Specificity

Cross-reactivity between the antibodies and any of the other analytes in this panel is non-detectable or negligible.

Application

  • Analytes: MMP-2, MMP-3, MMP-8, proMMP-9, MMP-12
  • Recommended Sample type: cell/tissue culture supernatant or extracts
  • Research Category: Inflammation & Immunology

Features and Benefits

Design your multiplex kit by choosing available analytes within this panel.

Storage and Stability

Recommended storage for kit components is 2 - 8°C.

Other Notes

Sensitivity: See protocol for sensitivities of individual analytes

Legal Information

MILLIPLEX is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Aquatic Chronic 2 - Eye Dam. 1 - Skin Sens. 1 - STOT RE 2

Target Organs

Respiratory Tract

Storage Class Code

10 - Combustible liquids


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Lucy M Hinder et al.
Experimental neurology, 305, 33-43 (2018-03-20)
Diabetic peripheral neuropathy is the most common complication of diabetes and a source of considerable morbidity. Numerous molecular pathways are linked to neuropathic progression, but it is unclear whether these pathways are altered throughout the course of disease. Moreover, the
Delphine Le Guennec et al.
Cancers, 14(1) (2022-01-12)
Our goal was to evaluate the effect of spontaneous physical activity on tumour immunity during aging. Elderly (n = 10/group, 33 weeks) ovariectomized C57BL/6J mice fed a hyperlipidic diet were housed in standard (SE) or enriched (EE) environments. After 4
Phillipe D O'Brien et al.
Neurobiology of disease, 73, 348-355 (2014-12-03)
Given the lack of treatments for diabetic neuropathy (DN), a common diabetic complication, accurate disease models are necessary. Characterization of the leptin-deficient BTBR ob/ob mouse, a type 2 diabetes model, demonstrated that the mice develop robust diabetes coincident with severe
Taotao Lao et al.
Proceedings of the National Academy of Sciences of the United States of America, 113(32), E4681-E4687 (2016-07-23)
Genetic variants in Hedgehog interacting protein (HHIP) have consistently been associated with the susceptibility to develop chronic obstructive pulmonary disease and pulmonary function levels, including the forced expiratory volume in 1 s (FEV1), in general population samples by genome-wide association
Ludovic Boytard et al.
Nature communications, 11(1), 4311-4311 (2020-08-29)
Pulmonary disease increases the risk of developing abdominal aortic aneurysms (AAA). However, the mechanism underlying the pathological dialogue between the lungs and aorta is undefined. Here, we find that inflicting acute lung injury (ALI) to mice doubles their incidence of

Related Content

Immunology multiplex assays, like MILLIPLEX® assays, allow researchers to investigate more immune biomarkers simultaneously, saving time and resources. This is especially critical to immunology research, where numerous analytes play a role in different pathways.

Immunology multiplex assays, like MILLIPLEX® assays, allow researchers to investigate more immune biomarkers simultaneously, saving time and resources. This is especially critical to immunology research, where numerous analytes play a role in different pathways.

Immunology multiplex assays, like MILLIPLEX® assays, allow researchers to investigate more immune biomarkers simultaneously, saving time and resources. This is especially critical to immunology research, where numerous analytes play a role in different pathways.

Immunology multiplex assays, like MILLIPLEX® assays, allow researchers to investigate more immune biomarkers simultaneously, saving time and resources. This is especially critical to immunology research, where numerous analytes play a role in different pathways.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service