Skip to Content
Merck
All Photos(4)

Documents

MABT837

Sigma-Aldrich

Anti-LBPA Antibody, clone 6C4

clone 6C4, from mouse

Synonym(s):

LBPA

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

mouse

Quality Level

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

6C4, monoclonal

species reactivity (predicted by homology)

all

technique(s)

ELISA: suitable
dot blot: suitable
electron microscopy: suitable
immunocytochemistry: suitable

isotype

IgG1κ

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

bacterial ... Lbpa(61281717)

General description

Lyso bis-phosphatidic acid (LBPA, Lysobisphosphatidic acid, L(bis)PA) is a lipid that is exclusively located in late endosomes and is used as a late endosome marker. One of the characteristic features of late endosomes is a complex system of internal membranes within the lumen that contains large amounts of the unique, poorly degradable LBPA, and thus forms a specialized membrane domain within endosomes. Late endosomes function not only as a major protein-sorting compartment, but also an obligatory station for LDL and other endocytosed ligands destined for degradation. Research shows that the LBPA-rich membranes within late endosomes regulate cholesterol transport, presumably by acting as a collection and distribution device. The genetic disease Niemann–Pick type C (NPC) is characterized by both lysosomal and endosomal storage disorders. In skin fibroblasts from NPC patients, cholesterol accumulation is seen within vesicles containing late endosome markers LBPA and Rab7.

Specificity

Clone 6C4 specifically detects LBPA, but not PC, PE, SM, PS, CL, PI, Sul, Chol, Cer, or CE purified from baby hamster kidney (BHK) cell lipid extract (Kobayahsi, T., et al. (1998). Nature. 392(6672):193-197).
Target molecule is not species-specific.

Immunogen

Endosomal membranes from baby hamster kidney fibroblasts (BHK) corresponding to Hamster LBPA.

Application

Immunocytochemistry Analysis: 4.0 µg/mL from a representative lot detected LBPA in A431, HUVEC and NIH/3T3 cells.
Immunocytochemistry Analysis: A representative lot detected late endosome LBPA immunoreactivity by fluorescent immunocytochemistry using MLN64-overexpressing MCF7 cells (Alpy, F., et al. (2001). J Biol Chem. 276(6):4261-4269).
Immunocytochemistry Analysis: A representative lot detected late endosome LBPA immunoreactivity in skin fibroblasts from healthy donors, as well as Niemann–Pick type C (NPC) and Tay-Sachs disease patients by fluorescent immunocytochemistry (Kobayahsi, T., et al. (1999). Nat Cell Biol. 1(2):113-118).
Immunocytochemistry Analysis: A representative lot detected late endosome LBPA immunoreactivity in baby hamster kidney (BHK) cells by fluorescent immunocytochemistry (Kobayahsi, T., et al. (1998). Nature. 392(6672):193-197).
Electron Microscopy Analysis: A representative lot detected similar cellular LBPA distribution in the skin fibroblasts from autosomal recessive Niemann–Pick type C disease (NPC) patients as seen in baby hamster kidney (BHK) cells (Kobayahsi, T., et al. (1999). Nat Cell Biol. 1(2):113-118).
Electron Microscopy Analysis: A representative lot detected LBPA immunoreactivity specifically associated with the late endosomes in baby hamster kidney (BHK) cells, but not the lgp120-positive limiting membranes of the same endosomes (Kobayahsi, T., et al. (1998). Nature. 392(6672):193-197).
ELISA Analysis: A representative lot specifically detected wells coated with total baby hamster kidney (BHK) cell lipid extract or purified LBPA, but not other lipids (PC, PE, SM, PS, CL, or PI) purified BHK lipid extract (Kobayahsi, T., et al. (1998). Nature. 392(6672):193-197)
Dot Blot Analysis: A representative lot specifically detected purified LBPA spotted on a HPTLC plate, but not other lipids (PC, PE, SM, PS, CL, PI, Sul, Chol, Cer, CE) purified from baby hamster kidney (BHK) cell lipid extract (Kobayahsi, T., et al. (1998). Nature. 392(6672):193-197).
Research Category
Cell Structure
Research Sub Category
Adhesion (CAMs)
This Anti-LBPA Antibody, clone 6C4 is validated for use in Immunocytochemistry, Electron Microscopy, ELISA and Dot Blot for the detection of LBPA.

Quality

Evaluated by Immunocytochemistry in HeLa cells.

Immunocytochemistry Analysis: 4.0 µg/mL of this antibody stained late endosomes in HeLa cells.

Physical form

Format: Purified
Protein G Purified
Purified mouse monoclonal IgG1κ antibody in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Other Notes

Concentration: Please refer to lot specific datasheet.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jessica Venugopal et al.
Scientific reports, 12(1), 16437-16437 (2022-10-01)
Sickle cell disease (SCD) is associated with altered plasma and erythrocyte lipid profiles. In a previous study, SCD mice with deficiency of proprotein convertase subtilisin/kexin type 9 (PCSK9) were observed to have more severe anemia and increased sickling compared to control
Curtis B Read et al.
mBio, 13(6), e0296122-e0296122 (2022-11-22)
Anaplasma phagocytophilum is the etiologic agent of the emerging infection, granulocytic anaplasmosis. This obligate intracellular bacterium lives in a host cell-derived vacuole that receives membrane traffic from multiple organelles to fuel its proliferation and from which it must ultimately exit
Pia Hartwig et al.
International journal of molecular sciences, 22(13) (2021-07-03)
For many years, the biology of glycosphingolipids was elucidated with the help of glucosylceramide synthase (GCS) inhibitors such as 1-phenyl-2-decanoylamino-3-morpholino-1-propanol (PDMP). Additionally, PDMP gained interest because of its chemosensitizing effects. Several studies have successfully combined PDMP and anti-cancer drugs in
Simon Zenke et al.
Nature communications, 13(1), 6459-6459 (2022-10-31)
Intercellular communication is crucial for collective regulation of cellular behaviors. While clustering T cells have been shown to mutually control the production of key communication signals, it is unclear whether they also jointly regulate their availability and degradation. Here we
Thach Tuan Pham et al.
Journal of extracellular vesicles, 12(8), e12354-e12354 (2023-08-09)
Extracellular vesicles (EVs) can be produced from red blood cells (RBCs) on a large scale and used to deliver therapeutic payloads efficiently. However, not much is known about the native biological properties of RBCEVs. Here, we demonstrate that RBCEVs are

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service