Skip to Content
Merck
All Photos(1)

Key Documents

MABS61

Sigma-Aldrich

Anti-PARG Antibody, clone D8B10

clone D8B10, from mouse

Synonym(s):

poly (ADP-ribose) glycohydrolase, poly(ADP-ribose) glycohydrolase 60 kDa isoform

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

mouse

Quality Level

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

D8B10, monoclonal

species reactivity

human

technique(s)

immunoprecipitation (IP): suitable
western blot: suitable

isotype

IgG2aκ

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... PARG(8505)

General description

Poly(ADP-ribose) glycohydrolase (PARG) is an enzyme possessing both endo- and exoglycosidase activity against poly (ADP-ribose) (PARP), rapidly degrading PARP to release large quantities of free ADP-ribose. PARG is involved in several cellular processes including; apoptosis, DNA repair, cell cycle progression, cell survival and cellular differentiation. During apoptosis, PARG is cleaved by caspase-3 suggesting that PARG activity is regulated during this process. Although encoded by one gene, PARG is present in different cellular localizations as different isoforms: Isoform 1 (111 kDa) is present in the nucleus, Isoform 2 (102 kDA) is in the cytoplasm and Isoform 3 (99 kDa) is mitochondrial. Studies have indicated a critical role for PARG isoform 1 in the quality of sperm chromatin, and subsequent embryonic survival.

Immunogen

Epitope: Unknown
Histidine-tagged recombinant protein corresponding to human PARG.

Application

Anti-PARG Antibody, clone D8B10 is an antibody against PARG for use in WB & IP.
Research Category
Signaling

Epigenetics & Nuclear Function
Research Sub Category
Hormones & Receptors

Chromatin Biology

Quality

Evaluated by Western Blot in Jurkat cell lysate.

Western Blot Analysis: 0.5 µg/mL of this antibody detected PARG on 10 µg of Jurkat cell lysate.

Target description

~ 102 kDa observed. Other isoforms may be observed in some lysates at 111 and 99 kDa

Physical form

Format: Purified
Protein G
Purified mouse monoclonal IgG2aκ in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Analysis Note

Control
Jurkat cell lysate

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Kaede Tsuda et al.
Cancer reports (Hoboken, N.J.), 6(2), e1709-e1709 (2022-09-03)
Poly(ADP-ribose) glycohydrolase (PARG) is a key enzyme in poly(ADP-ribose) (PAR) metabolism and a potential anticancer target. Many drug candidates have been developed to inhibit its enzymatic activity. Additionally, PDD00017273 is an effective and selective inhibitor of PARG at the first
Jean-Christophe Amé et al.
Methods in molecular biology (Clifton, N.J.), 1608, 395-413 (2017-07-12)
The purification of Poly(ADP-ribose) glycohydrolase (PARG) from overexpressing bacteria Escherichia coli is described here to a fast and reproducible one chromatographic step protocol. After cell lysis, GST-PARG-fusion proteins from the crude extract are affinity purified by a Glutathione 4B Sepharose
Yajie Zhang et al.
Nature methods, 10(10), 981-984 (2013-08-21)
Poly(ADP-ribosyl)ation is catalyzed by a family of enzymes known as PARPs. We describe a method to characterize the human aspartic acid- and glutamic acid-ADP-ribosylated proteome. We identified 1,048 ADP-ribosylation sites on 340 proteins involved in a wide array of nuclear
Xiaoxing Feng et al.
Molecular cancer, 11, 48-48 (2012-07-31)
Cell death induced by poly(ADP-ribose) (PAR) and mediated by apoptosis-inducing factor (AIF) is well-characterized in models of ischemic tissue injury, but their roles in cancer cell death after chemotherapy are less understood. Here we investigated the roles of PAR and
Yukihide Watanabe et al.
The Journal of biological chemistry, 291(24), 12706-12723 (2016-04-30)
We previously established a mechanism of negative regulation of transforming growth factor β signaling mediated by the nuclear ADP-ribosylating enzyme poly-(ADP-ribose) polymerase 1 (PARP1) and the deribosylating enzyme poly-(ADP-ribose) glycohydrolase (PARG), which dynamically regulate ADP-ribosylation of Smad3 and Smad4, two

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service