Skip to Content
Merck
All Photos(2)

Key Documents

MABS193

Sigma-Aldrich

Anti-v-Src Antibody, clone 327

clone 327, from mouse

Synonym(s):

Proto-oncogene tyrosine-protein kinase Src, Proto-oncogene c-Src, pp60c-src, p60-Src

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

mouse

Quality Level

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

327, monoclonal

species reactivity

mouse

technique(s)

western blot: suitable

isotype

IgG1κ

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... SRC(6714)

General description

v-Src is the product of the Rous sarcoma virus (RSV) oncogene, and functions as a non-receptor tyrosine kinase belonging to the family of Src proteins. Srcs are typically activated by dimerized cell surface receptors such as integrins, receptor tyrosine kinases, and cytokine receptors. Activated Src kinases phosphorylate the cytoplasmic domains of these receptors enabling interaction between these receptors and their downstream effector molecules. Src proteins are involved in many cellular processs such as apoptosis, immunity, gene expression, cell adhesion, cell migration, and cell transformation. Previous studies have reported that v-Src plays an important role in tumorigenesis.

Immunogen

Recombinant protein corresponding to purified pp60src from bacterial recombinants which direct the synthesis of v-Src.

Application

Anti-v-Src Antibody, clone 327 detects level of v-Src & has been published & validated for use in Western Blotting.
Research Category
Signaling
Research Sub Category
Immunological Signaling

Quality

Evaluated by Western Blot in NIH/3T3 cell lysate.

Western Blot Analysis: 0.1 µg/mL of this antibody detected v-Src in 10 µg of NIH/3T3 cell lysate.

Target description

~60 kDa observed

Physical form

Format: Purified
Protein G Purified
Purified mouse monoclonal IgG1κ in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Analysis Note

Control
NIH/3T3 cell lysate

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Lilian Varricchio et al.
Molecular cancer research : MCR, 5(11), 1213-1221 (2007-11-21)
This report offers direct evidence that association of the estradiol receptor (ER) with Src triggered by steroid agonists or growth factors controls breast and prostate cancer cell growth. This association is abolished in whole cells and in vitro by a
Hila Toledano-Katchalski et al.
Molecular and cellular biology, 23(15), 5460-5471 (2003-07-16)
cyt-PTP epsilon is a naturally occurring nonreceptor form of the receptor-type protein tyrosine phosphatase (PTP) epsilon. As such, cyt-PTP epsilon enables analysis of phosphatase regulation in the absence of extracellular domains, which participate in dimerization and inactivation of the receptor-type
Matthew H Herynk et al.
Molecular cancer therapeutics, 5(12), 3023-3031 (2006-12-19)
It has long been appreciated that estrogenic signaling contributes to breast cancer progression. c-Src is also required for a number of processes involved in tumor progression and metastasis. We have previously identified the K303R mutant estrogen receptor alpha (ERalpha) that
Eynat Finkelshtein et al.
Molecular biology of the cell, 25(11), 1808-1818 (2014-04-04)
Female mice lacking protein tyrosine phosphatase ε (PTP ε) are mildly osteopetrotic. Osteoclasts from these mice resorb bone matrix poorly, and the structure, stability, and cellular organization of their podosomal adhesion structures are abnormal. Here we compare the role of
Gabriella Castoria et al.
PloS one, 8(10), e76899-e76899 (2013-10-17)
Hormones and growth factors influence the proliferation and invasiveness of human mesenchymal tumors. The highly aggressive human fibrosarcoma HT1080 cell line harbors classical androgen receptor (AR) that responds to androgens triggering cell migration in the absence of significant mitogenesis. As

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service