Skip to Content
Merck
All Photos(1)

Key Documents

MABN59

Sigma-Aldrich

Anti-ACE2 Antibody, clone 4G5.1

clone 4G5.1, from mouse

Synonym(s):

Angiotensin-converting enzyme 2, ACE-related carboxypeptidase, Angiotensin-converting enzyme homolog, ACEH, Metalloprotease MPROT15

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41
tag:
GST tagged
application:
WB
clone:
4G5.1, monoclonal
species reactivity:
human
citations:
10
technique(s):
western blot: suitable

biological source

mouse

Quality Level

tag

GST tagged

antibody form

purified antibody (protein G)

antibody product type

primary antibodies

clone

4G5.1, monoclonal

mol wt

~62 kDa

species reactivity

human

technique(s)

western blot: suitable

isotype

IgG1κ

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

2-8°C

target post-translational modification

unmodified

Gene Information

human ... ACE2(59272)

General description

Angiotensin-converting enzyme 2 (ACE2) is a transmembrane protein belonging to the metallopeptidases (M2) family. It is a membrane-associated carboxypeptidase, expressed in the vascular endothelial cells of the heart, kidney, testis, gastrointestinal system and in the epithelial cells of lungs. ACE2 consists of an N-terminal peptidase domain (PD) and a C-terminal collectrin-like domain (CLD). It also comprises a single catalytic domain that is similar to that of ACE. The gene ACE2 is mapped to human chromosome location Xp22.2. ACE2 is a structural homolog ACE.

Specificity

This antibody recognizes ACE2.

Immunogen

GST-tagged recombinant protein corresponding to human ACE2.

Application

Anti-ACE2 Antibody, clone 4G5.1 detects level of ACE2 & has been validated for use in Western Blotting.
ELISA Analysis: A representative lot detected ACE2 in a direct ELISA.
Research Category
Signaling
Research Sub Category
Signaling Neuroscience

Biochem/physiol Actions

Angiotensin-converting enzyme-2 (ACE2) produces angiotensin (1-9) by hydrolysis of angiotensin 1. Angiotensin (1-9) is a precursor for angiotensin (1-7). ACE2 may also directly produce angiotensin (1-7) by conversion of angiotensin II, which is involved in vasoconstriction.(2) Angiotensin (1-7) plays an important role in the renin-angiotensin system (RAS) signaling pathway and is involved in bradykinin-induced vasodilation. Hence, ACE2 antagonizes the effects of its homolog, ACE. ACE2 is insensitive to commonly used inhibitors such as lisinopril and captopril. This gene is upregulated in heart failure due to the overactivity of angiotensin II. ACE2 is studied for its potential therapeutic value for hypertension and heart failure. It binds to the viral envelope protein spike (S) protein and acts as a major entry receptor for severe acute respiratory syndrome coronavirus (SARS-CoV) and SARS-CoV-2 in human cells. Hence, it is considered as a potential therapeutic target for COVID-19. ACE2 exhibits protection from lung injury.

Quality

Evaluated by Western Blot in HEK293 cell lysate.

Western Blot Analysis: A 1:2,000 dilution of this antibody detected ACE2 in 10 µg of HEK293 cell lysate.

Target description

~62 kDa observed. Uniport describes 2 isoforms at ~92 kDa (Isoform 1) and ~64 kDa (Isoform 2) due to alternative splicing.

Linkage

Replaces: MAB5676

Physical form

Format: Purified
Protein G Purified
Purified mouse monoclonal IgG1κ in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Analysis Note

Control
HEK293 cell lysate

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Yuebo Zhang et al.
Frontiers in physiology, 9, 1370-1370 (2018-10-27)
Body fat distribution contributes to obesity-related metabolic and cardiovascular disorders. Visceral fat is more detrimental than subcutaneous fat. However, the mechanisms underlying visceral fat-mediated cardiometabolic dysregulation are not completely understood. Localized increases in expression of the renin angiotensin system (RAS)
Ace revisited: a new target for structure-based drug design.
K Ravi Acharya et al.
Nature reviews. Drug discovery, 2(11), 891-902 (2003-12-12)
Angiotensin-converting enzyme 2 (ACE2) as a SARS-CoV-2 receptor: molecular mechanisms and potential therapeutic target.
Haibo Zhang et al.
Intensive care medicine, 46(4), 586-590 (2020-03-04)
Andrew B Goulter et al.
BMC medicine, 2, 19-19 (2004-05-21)
ACE2 is a novel homologue of angiotensin converting enzyme (ACE). ACE2 is highly expressed in human heart and animal data suggest that ACE2 is an essential regulator of cardiac function in vivo. Since overactivity of the renin-angiotensin system contributes to
Mir S Adil et al.
Drug discoveries & therapeutics, 14(5), 256-258 (2020-10-30)
In the ongoing coronavirus diseases-2019 (COVID-19) crisis that caused immense suffering and deaths, the choice of therapy for the prevention and life-saving conditions must be based on sound scientific evidence. Uncertainty and apprehension are exacerbated in people using angiotensin-converting enzyme

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service