MABF366
Anti-CD11b Antibody (human/mouse), APC, clone M1/70
clone M1/70, 0.2 mg/mL, from rat
Synonym(s):
Integrin alpha-M, CD11 antigen-like family member B, CR-3 alpha chain, Cell surface glycoprotein MAC-1 subunit alpha, Leukocyte adhesion receptor MO1, Neutrophil adherence receptor, CD antigen CD11b
About This Item
Recommended Products
biological source
rat
Quality Level
conjugate
Allophycocyanin conjugate
antibody form
purified antibody
antibody product type
primary antibodies
clone
M1/70, monoclonal
species reactivity
mouse, human
concentration
0.2 mg/mL
technique(s)
flow cytometry: suitable
immunofluorescence: suitable
immunohistochemistry: suitable
immunoprecipitation (IP): suitable
isotype
IgG2bκ
shipped in
wet ice
target post-translational modification
unmodified
Gene Information
human ... ITGAM(3684)
Related Categories
General description
Immunogen
Application
Inflammation & Immunology
Physical form
Storage and Stability
Disclaimer
Not finding the right product?
Try our Product Selector Tool.
Storage Class Code
12 - Non Combustible Liquids
WGK
nwg
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service