Skip to Content
Merck
All Photos(1)

Key Documents

MABF120

Sigma-Aldrich

Anti-EGFR Antibody, clone 225 (Azide-free)

clone 225, from mouse

Synonym(s):

Epidermal growth factor receptor, Proto-oncogene c-ErbB-1, Receptor tyrosine-protein kinase erbB-1

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

mouse

Quality Level

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

225, monoclonal

species reactivity

human

technique(s)

immunoprecipitation (IP): suitable
neutralization: suitable

isotype

IgG1κ

NCBI accession no.

UniProt accession no.

shipped in

dry ice

target post-translational modification

unmodified

Gene Information

human ... EGFR(1956)

General description

The epidermal growth factor receptor (EGFR; Proto-oncogene c-ErbB-1) is a ubiquitously receptor tyrosine kinase that is activated by a variety of ligands including EGF, EPGN, TGFA, and EREG. Ligand-bound EGFR undergoes dimerization and autophosphorylation exposing cytoplasmic domains which allow interaction with adaptor proteins and induction of several signaling pathways including PI3-AKT; PLCγ-PKC; Notch1; and Ras-Raf. EGFR therefore contributes to a diverse range of cellular processes inlcuding cell proliferation. Several studies have reported that EGFR signaling plays an important role in the development of various tumors. EGFR is regulated by PTPRJ and PTPRK phosphatases, and by the endocytic machinery including EPS15 that mediates internalization and degradation of the receptor.

Immunogen

Partially purified human EGFR from A431 cells

Application

Anti-EGFR Antibody, clone 225 (Azide-free) is an antibody against EGFR for use in Immunoprecipitation, Neutralizing.
Neutralizing Assay Analysis: A representative lot was used by an independent laboratory in A431 cells. (Kawamoto, T., et al. (1984). Journal of Biological Chemistry. 259(12):7761-7766.)

Quality

Evaluated by Immunoprecipitation in A431 cell lysate.

Immunoprecipitation Analysis: 10 µg of this antibody immunoprecipitated EGFR from A431 cell lysate.

Target description

~170 kDa observed. Uniprot gives a calculated molecular weight of 135 but can be observed at ~170 kDa due to phosphorylation. (Konishi, A., et al. (2003). The Journal of Biological Chemistry. 278(37):35049–35056.)

Linkage

Replaces: 04-338

Physical form

Format: Purified

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Eduardo Torre et al.
Cell systems, 6(2), 171-179 (2018-02-20)
Although single-cell RNA sequencing can reliably detect large-scale transcriptional programs, it is unclear whether it accurately captures the behavior of individual genes, especially those that express only in rare cells. Here, we use single-molecule RNA fluorescence in situ hybridization as
Sabrina A Maisel et al.
Journal of translational medicine, 17(1), 201-201 (2019-06-20)
The human epidermal growth factor receptor (HER) family of transmembrane tyrosine kinases is overexpressed and correlates with poor prognosis and decreased survival in many cancers. The receptor family has been therapeutically targeted, yet tyrosine kinase inhibitors (TKIs) do not inhibit
Tsuyoshi Oguma et al.
Journal of immunology (Baltimore, Md. : 1950), 187(2), 999-1005 (2011-06-21)
Allergic bronchopulmonary mycosis, characterized by excessive mucus secretion, airflow limitation, bronchiectasis, and peripheral blood eosinophilia, is predominantly caused by a fungal pathogen, Aspergillus fumigatus. Using DNA microarray analysis of NCI-H292 cells, a human bronchial epithelial cell line, stimulated with fungal
Jason Buehler et al.
PLoS pathogens, 12(5), e1005655-e1005655 (2016-05-25)
Herpesviruses persist indefinitely in their host through complex and poorly defined interactions that mediate latent, chronic or productive states of infection. Human cytomegalovirus (CMV or HCMV), a ubiquitous β-herpesvirus, coordinates the expression of two viral genes, UL135 and UL138, which
Sydney M Shaffer et al.
Cell, 182(4), 947-959 (2020-08-01)
Non-genetic factors can cause individual cells to fluctuate substantially in gene expression levels over time. It remains unclear whether these fluctuations can persist for much longer than the time of one cell division. Current methods for measuring gene expression in

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service