Skip to Content
Merck
All Photos(1)

Key Documents

MABE302

Sigma-Aldrich

Anti-2,2,7-Trimethylguanosine Antibody, clone K121

clone K121, from mouse

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

mouse

Quality Level

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

K121, monoclonal

species reactivity

human, mouse

technique(s)

affinity chromatography: suitable
immunocytochemistry: suitable
immunoprecipitation (IP): suitable

isotype

IgG1κ

shipped in

wet ice

target post-translational modification

unmodified

General description

The 2,2,7-trimethylguanosine cap structure is found on the 5’ end of small nuclear RNAs and small nucleolar RNAs. Initially, these RNA molecules are capped with 7-monomethylguanosine (m7G) which is then methylated by the trimethylguanosine synthase enzyme to form 2,2,7-trimethylguanosine. The synthesis of 2,2,7-trimethylguanosine occurs in the cytoplasm and is an important marker for nuclear localization of RNA molecules. Capping of RNAs is an important requirement for RNA transport and splicing processes. Uncapped RNAs are rapidly degraded by the 5′ exoribonuclease enzyme. 2,2,7-trimethylguanosine capping may play a role in the expression of viral RNAs.

Application

Anti-2, 2, 7-Trimethylguanosine Antibody, clone K121 is a high quality Mouse Monoclonal Antibody for the detection of 2, 2, 7-Trimethylguanosine & has been validated in ICC, PAI & IP.
Immunoaffinity Purification Analysis: A representative lot from an independent laboratory was used in IAP (Krainer, A.R., et al. (1988). Nucleic Acids Res. 16(20):9415-9429.).

Immunoprecipitation Analysis: A representative lot from an independent laboratory was immunoprecipitated in IP (Moketi, S., et al. (2002). Mol Cell. 10(3):599-609.).

Quality

Evaluated by Immunocytochemistry in NIH/3T3, HeLa, and A431 cells.

Immunocytochemistry Analysis: A 1:500 dilution of this antibody detected 2,2,7-trimethylguanosine in NIH/3T3, HeLa, and A431 cells.

Physical form

Format: Purified

Analysis Note

Control
NIH/3T3, HeLa, and A431 cells

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ranjodh Sandhu et al.
Cell research, 23(4), 537-551 (2013-03-13)
Telomerase is a ribonucleoprotein enzyme typically required for sustained cell proliferation. Although both telomerase activity and the telomerase catalytic protein component, TbTERT, have been identified in the eukaryotic pathogen Trypanosoma brucei, the RNA molecule that dictates telomere synthesis remains unknown.
Cyrille Girard et al.
Nucleic acids research, 34(10), 2925-2932 (2006-06-02)
Neuronal degeneration in spinal muscular atrophy (SMA) is caused by reduced expression of the survival of motor neuron (SMN) protein. The SMN protein is ubiquitously expressed and is present both in the cytoplasm and in the nucleus where it localizes
Magdalena Strzelecka et al.
Nucleus (Austin, Tex.), 1(1), 96-108 (2011-02-18)
The Cajal body (CB) is an evolutionarily conserved nuclear subcompartment, enriched in components of the RNA processing machinery. The composition and dynamics of CBs in cells of living organisms is not well understood. Here we establish the zebrafish embryo as
Simon Henriet et al.
Current biology : CB, 29(19), 3193-3199 (2019-09-24)
An overwhelming majority of eukaryotic introns have GT/AG ends, whose identities play a critical role for their recognition and removal by the U2 spliceosome, a well-conserved complex of protein and RNAs. Introns with other splice sites exist at very low
Cristina Moreno-Castro et al.
Journal of cell science, 132(22) (2019-10-23)
Cajal bodies are nuclear organelles involved in the nuclear phase of small nuclear ribonucleoprotein (snRNP) biogenesis. In this study, we identified the splicing factor TCERG1 as a coilin-associated factor that is essential for Cajal body integrity. Knockdown of TCERG1 disrupts

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service