Skip to Content
Merck
All Photos(1)

Documents

MABC547

Sigma-Aldrich

Anti-Poly ADP-ribose Antibody, clone 10H

clone 10H, from mouse

Synonym(s):

Anti-PAR antibody

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

mouse

Quality Level

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

10H, monoclonal

species reactivity

human

technique(s)

immunofluorescence: suitable
immunoprecipitation (IP): suitable
western blot: suitable

isotype

IgG3κ

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... PARP1(142)

General description

Poly(ADP-ribose) polymerase is an abundant nuclear enzyme that catalyses the synthesis of poly(ADP-ribose) from nicotinamide adenine dinucleotide (NAD+). Poly(ADP-ribose) has an N-terminal DNA-binding domain containing two zinc-fingers, which is linked to the C-terminal NAD+-binding domain by a short region containing several glutamic acid residues that are sites of auto-poly (ADP-ribosyl) ation. Production of poly(ADP-ribose) within the cell is initiated by agents that generate DNA strand interruptions. The branched homopolymer chains may reach a length of 200–300 residues but are rapidly degraded after synthesis. The function of poly(ADP-ribose) synthesis is not clear, although it seems to be required for DNA repair.

Immunogen

Corresponding to human Poly ADP-ribose chain.

Application

Detect PARP using this mouse monoclonal antibody, Anti-Poly ADP-ribose Antibody, clone 10H validated for use in western blotting, IP & Immunofluorescence.
Immunoprecipitation Analysis: A representative lot was used by an an independent laboratory to immunoprecipitate Poly ADP-ribose in cultured supernatents from mouse erythrocytes coated with Poly ADP-ribose (Kawamitsu, H., et al. (1984). American Chemical Society. 3771-3777).
Immunofluorescence Analysis: A representative lot was used by an an independent laboratory to detect Poly (ADP-ribose) synthesis on transfected CV-1 cells via indirect immunofluorescence (J.H. Kupper, et al. J. Biol. Chem. (1990) 265:18721).
Research Category
Apoptosis & Cancer
Research Sub Category
Apoptosis - Additional

Quality

Evaluated by Western Blotting in untreated and UV treated HeLa cells.

Western Blotting Analysis: 1.0 µg/mL of this antibody detected UV treated HeLa cells. Little or no signal observed in untreated HeLa cells.

Target description

Observed molecular weight is the result of poly(ADP-ribose) polymer on protein receptors (such as histones and transcription factors).

Linkage

Replaces: MAB3192

Physical form

Format: Purified
Protein G Purified
Purified mouse monoclonal IgG3κ in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Yousef M O Alhammad et al.
bioRxiv : the preprint server for biology (2020-06-09)
Severe acute respiratory syndrome coronavirus 2 (SARS-CoV-2) and other SARS-like-CoVs encode 3 tandem macrodomains within non-structural protein 3 (nsp3). The first macrodomain, Mac1, is conserved throughout CoVs, and binds to and hydrolyzes mono-ADP-ribose (MAR) from target proteins. Mac1 likely counters
Jing Xu et al.
Cancer chemotherapy and pharmacology, 89(5), 683-695 (2022-04-15)
Although the use of PARP inhibitor has received considerable amount of attention in ovarian cancer, PARP inhibitor resistance still emerges with disease progression. PI3K/AKT pathway inhibitors have been proposed to synergize with PARP inhibition to slow tumor growth, but the
Umeshkumar Vekariya et al.
Nature communications, 15(1), 5822-5822 (2024-07-11)
DNA polymerase theta (Polθ)-mediated end-joining (TMEJ) repairs DNA double-strand breaks and confers resistance to genotoxic agents. How Polθ is regulated at the molecular level to exert TMEJ remains poorly characterized. We find that Polθ interacts with and is PARylated by
Lin Liu et al.
Science advances, 8(19), eabh2332-eabh2332 (2022-05-12)
Tumor necrosis factor (TNF) is a key component of the innate immune response. Upon binding to its receptor, TNFR1, it promotes production of other cytokines via a membrane-bound complex 1 or induces cell death via a cytosolic complex 2. To
Song My Hoang et al.
Nature structural & molecular biology, 27(12), 1152-1164 (2020-10-14)
The synthesis of poly(ADP-ribose) (PAR) reconfigures the local chromatin environment and recruits DNA-repair complexes to damaged chromatin. PAR degradation by poly(ADP-ribose) glycohydrolase (PARG) is essential for progression and completion of DNA repair. Here, we show that inhibition of PARG disrupts

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service