MAB8910
Anti-Measles Blend Antibody, matrix protein, clones CV8, CV9
Chemicon®, from mouse
Synonym(s):
Rubeola
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41
Recommended Products
biological source
mouse
Quality Level
antibody form
purified antibody
antibody product type
primary antibodies
clone
CV8, monoclonal
CV9, monoclonal
species reactivity
human
manufacturer/tradename
Chemicon®
technique(s)
immunofluorescence: suitable
isotype
IgG
shipped in
wet ice
Specificity
Recognizes the measles matrix protein by indirect immunofluorescence.
Immunogen
Epitope: matrix protein
Application
Detect Measles Blend using this Anti-Measles Blend Antibody, matrix protein, clones CV8, CV9 validated for use in IF.
Indirect Immunofluorescence
Final working dilutions must be determined by end user.
Final working dilutions must be determined by end user.
Research Category
Infectious Diseases
Infectious Diseases
Research Sub Category
Infectious Diseases - Viral
Infectious Diseases - Viral
Physical form
Format: Purified
In 0.02M PB with 0.25M NaCl containing 0.1% sodium azide, pH 7.6.
Storage and Stability
Maintain at 2-8°C in aliquots for up to 12 months.
Other Notes
Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.
Legal Information
CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany
Disclaimer
Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.
Not finding the right product?
Try our Product Selector Tool.
Storage Class Code
10 - Combustible liquids
WGK
WGK 2
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
RNA interference with measles virus N, P, and L mRNAs efficiently prevents and with matrix protein mRNA enhances viral transcription.
Reuter, T; Weissbrich, B; Schneider-Schaulies, S; Schneider-Schaulies, J
Journal of virology null
Measles virus nucleocapsid transport to the plasma membrane requires stable expression and surface accumulation of the viral matrix protein.
Nicole Runkler,Christine Pohl,Sibylle Schneider-Schaulies,Hans-Dieter Klenk,Andrea Maisner
Cellular Microbiology null
Measles virus M and F proteins associate with detergent-resistant membrane fractions and promote formation of virus-like particles.
Pohl, C; Duprex, WP; Krohne, G; Rima, BK; Schneider-Schaulies, S
The Journal of General Virology null
Measles virus M protein driven particle production does not involve the ESCRT system.
A Salditt, S Koethe, C Pohl, H Harms, L Kolesnikova, S Becker, S Schneider-Schaulies
The Journal of General Virology null
Hasan Kweder et al.
Advances in virology, 2015, 769837-769837 (2015-11-21)
Subacute Sclerosing Panencephalitis (SSPE), a rare lethal disease of children and young adults due to persistence of measles virus (MeV) in the brain, is caused by wild type (wt) MeV. Why MeV vaccine strains never cause SSPE is completely unknown.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service