Skip to Content
Merck
All Photos(1)

Key Documents

MAB8616

Sigma-Aldrich

Anti-Varicella-Zoster Virus Antibody, Immediate Early Gene 62

Chemicon®, from mouse

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

mouse

Quality Level

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

monoclonal

species reactivity

human

manufacturer/tradename

Chemicon®

technique(s)

immunofluorescence: suitable
immunohistochemistry: suitable
immunoprecipitation (IP): suitable
western blot: suitable

isotype

IgG1

shipped in

wet ice

Specificity

Recognizes the VZV immediate early gene 62. No cross-reactivity to HSV.

Immunogen

Epitope: Immediate Early Gene 62
VZV HZ strain.

Application

Indirect immunofluorescence: shows a nuclear and cytoplasmic staining pattern.

Immunoprecipitation

Western blotting

Final working dilutions must be determined by end user.
Research Category
Infectious Diseases
Research Sub Category
Infectious Diseases - Viral
This Anti-Varicella-Zoster Virus Antibody, Immediate Early Gene 62 is validated for use in IF, IP, WB, IH for the detection of Varicella-Zoster Virus.

Physical form

Format: Purified
Purified immunoglobulin. In 0.02M PB with 0.25M NaCl, pH=7.6 with 0.1% Sodium azide.

Storage and Stability

Maintain at 2-8°C for up to 12 months.

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Legal Information

CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Myelin-associated glycoprotein mediates membrane fusion and entry of neurotropic herpesviruses.
Suenaga T, Satoh T, Somboonthum P, Kawaguchi Y, Mori Y, Arase H
Proceedings of the National Academy of Sciences of the USA null
Immunohistochemical detection of intra-neuronal VZV proteins in snap-frozen human ganglia is confounded by antibodies directed against blood group A1-associated antigens.
Werner J D Ouwendijk,Sarah E Flowerdew,Desiree Wick,Anja K E Horn,Inga Sinicina et al.
Journal of Neurovirology null
Identification of a varicella-zoster virus replication inhibitor that blocks capsid assembly by interacting with the floor domain of the major capsid protein.
Naoki Inoue,Misato Matsushita,Yoshiko Fukui,Souichi Yamada,Mihoko Tsuda,Chizuka Higashi et al.
Journal of virology null
Stefan L Oliver et al.
Journal of virology, 91(1) (2016-11-01)
The highly conserved herpesvirus glycoprotein complex gB/gH-gL mediates membrane fusion during virion entry and cell-cell fusion. Varicella-zoster virus (VZV) characteristically forms multinucleated cells, or syncytia, during the infection of human tissues, but little is known about this process. The cytoplasmic
Generation of a reporter cell line for detection of infectious varicella-zoster virus and its application to antiviral studies.
Wang, GQ; Suzutani, T; Yamamoto, Y; Fukui, Y; Nozawa, N; Schmid, DS; Kurane, I; Inoue, N
Antimicrobial agents and chemotherapy null

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service