Skip to Content
Merck
All Photos(5)

Key Documents

MAB5374

Sigma-Aldrich

Anti-Huntingtin Protein Antibody, clone mEM48

culture supernatant, clone mEM48, Chemicon®

Synonym(s):

Huntingtin

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

mouse

Quality Level

antibody form

culture supernatant

antibody product type

primary antibodies

clone

mEM48, monoclonal

species reactivity

human

species reactivity (predicted by homology)

mouse, rat

manufacturer/tradename

Chemicon®

technique(s)

immunocytochemistry: suitable
immunohistochemistry: suitable
western blot: suitable

isotype

IgG

NCBI accession no.

UniProt accession no.

target post-translational modification

unmodified

Gene Information

General description

Huntington disease (HD) is a hereditary, progressive, neurodegenerative ailment characterized by personality changes, motor impairment and subcortical dementia. The molecular basis of the disease involves the expansion of the trinucleotide CAG, coding for polyglutamine in the first exon of a chromosome four gene (4p16.3), which normally produces a widely expressed 3136 a.a. (~350 kDa) protein huntingtin with unclear function. The protein is found in the perinuclear region along with microtubules, and in the centrosomal region along with gamma-tubulin. Huntingtin is necessary for neuronal survival and is involved in synaptic vesicle trafficking, microtubule binding and may also have a role in apoptosis. In the HD condition, neuronal cells with the mutant form of huntingtin possess intranuclear aggregations of the N-terminal fragment, causing damaging inclusions in perinuclear locations and striatal neuron cell death. Wild-type huntington and anti-huntingtin reduce aggregation and cellular toxicity of the mutant huntingtin form in mammalian cell models of HD. Huntingtin is known to interact with GAPDH, HAP-1, SP1 and TAFII130.

Specificity

Reacts with human huntingtin protein (both native and recombinant protein). MAB5374 reacts with mutant huntingtin in patients and in transgenic animals that express different numbers of repeats (from 82 to 150 glutamines). Thus, it should recognize different forms of mutant huntingtin.

Immunogen

GST fusion protein from the first 256 amino acids from human huntingtin with the deletion of the polyglutamine tract.

Application

Detect Huntingtin Protein using this Anti-Huntingtin Protein Antibody, clone mEM48 validated for use in IC, IH & WB.
Immunohistochemistry:
1:50-1:100 of a previous lot using ABC on 4% paraformaldehyde fixed tissue. Suggested dilution buffer is PBS containing 3% BSA. The antibody works on paraffin embedded tissue sections.
Suggested dilution buffer is PBS containing 3% BSA. The antibody works on paraffin embedded tissue sections. Yu, Z et al (2002) Hum. Mole. Genetics 11(8):905-914. (http://hmg.oxfordjournals.org/cgi/content/full/11/8/905) for good IHC methods and photos of mEM48 on rodent tissues with human transgenic material.

Immunocytochemistry:
light 4% PFA fixation followed by 0.1% triton X-100 incubation prior to blocking is suggested.
A previous lot of this antibody was used in IC.

Western blot:
1:50-1:500 using ECL depending on the level of mutant protein. Suggested dilution buffer is PBS containing 3% BSA or PBS containing 5% non-fat milk.
Nuclear fraction preparations enhance signals; monomeric protein ~80kDa; aggregates are common which can be >200kDa in size.

Optimal working dilutions must be determined by the end user.
Research Category
Neuroscience
Research Sub Category
Neurodegenerative Diseases

Quality

Routinely evaluated by Western Blot on HEK293 lysates.

Western Blot Analysis:
1:1000 dilution of this lot detected Huntingtin Protein on 10 μg of HEK293 lysates.

Target description

over 200 kDa

Physical form

Culture Supernatant mouse monoclonal IgG containing no preservative.
Unpurified

Storage and Stability

Stable for 6 months at -20ºC in undiluted aliquots from date of receipt.
Handling Recommendations: Upon receipt, and prior to removing the cap, centrifuge the vial and gently mix the solution. Aliquot into microcentrifuge tubes and store at -20°C. Avoid repeated freeze/thaw cycles, which may damage IgG and affect product performance.

Analysis Note

Control
Normal human cerebral cortex lysate, mouse brain cortex samples from HD or wild type mice

HEK293 lysates.

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Legal Information

CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

An engineered viral protease exhibiting substrate specificity for a polyglutamine stretch prevents polyglutamine-induced neuronal cell death.
Sellamuthu, S; Shin, BH; Han, HE; Park, SM; Oh, HJ; Rho, SH; Lee, YJ; Park, WJ
Testing null
Inhibition of the striatal specific phosphodiesterase PDE10A ameliorates striatal and cortical pathology in R6/2 mouse model of Huntington's disease.
Giampa, C; Laurenti, D; Anzilotti, S; Bernardi, G; Menniti, FS; Fusco, FR
Testing null
Neuroprotective effects of inositol 1,4,5-trisphosphate receptor C-terminal fragment in a Huntington's disease mouse model.
Tang, TS; Guo, C; Wang, H; Chen, X; Bezprozvanny, I
The Journal of Neuroscience null
Tomomi Imamura et al.
Scientific reports, 6, 33861-33861 (2016-09-23)
We identified drug seeds for treating Huntington's disease (HD) by combining in vitro single molecule fluorescence spectroscopy, in silico molecular docking simulations, and in vivo fly and mouse HD models to screen for inhibitors of abnormal interactions between mutant Htt
Chun-Chieh Hsu et al.
Scientific reports, 6, 23387-23387 (2016-03-19)
Bacterial infection-induced sepsis is the leading cause of septic inflammatory disease. Rhodostomin (Rn), a snake venom disintegrin, was previously reported to interact with the αVβ3 integrin and the TLR4 on phagocyte in attenuating LPS-induced endotoxemia. In this report, we further

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service