Skip to Content
Merck
All Photos(1)

Key Documents

MAB3536

Sigma-Aldrich

Anti-Lamin B2 Antibody, clone LN43

clone LN43, Chemicon®, from mouse

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

mouse

Quality Level

antibody form

culture supernatant

antibody product type

primary antibodies

clone

LN43, monoclonal

species reactivity

mouse, hamster, human, Xenopus

manufacturer/tradename

Chemicon®

technique(s)

flow cytometry: suitable
immunohistochemistry: suitable
western blot: suitable

isotype

IgG1

NCBI accession no.

UniProt accession no.

shipped in

dry ice

target post-translational modification

unmodified

Gene Information

human ... LMNB2(84823)

General description

Nuclear lamins form a network of filaments at the nucleoplasmic site of the nuclear membrane. Two main subtypes of nuclear lamins can be distinguished: A-type lamins and B-type lamins. The A-type lamins comprise a set of three proteins arising from the same gene by alternative splicing, i.e. lamin A, lamin C and lamin Adel 10, while the B-type lamins include two proteins arising from two distinct genes, i.e. lamin B1 and lamin B2.

Specificity

Lamin B2. Recognizes an epitope located in the C-terminal part of lamin B2.

Immunogen

Detergent insoluble fraction of potoroo cell line PtK1

Application

Research Category
Cell Structure
Research Sub Category
Cytoskeleton
This Anti-Lamin B2 Antibody, clone LN43 is validated for use in FC, WB, IH for the detection of Lamin B2.
Western blotting: 1:100-1:1,000; nuclear preparations recommended; detects 67-68kDa lamin B2 protein

Immunocytochemistry on fixed cells (methanol/acetone fixation): 1:100-1:200.Flow cytometry: 1:100-1:200.

Optimal working dilutions must be determined by the end user.

Physical form

Format: Purified
Purified culture supernatant. Liquid in PBS containing 0.09% sodium azide.

Storage and Stability

Maintain -20°C in undiluted aliquots for up to 6 months. Avoid repeated freeze/thaw cycles.

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Legal Information

CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Lucia Devitt et al.
Cell communication and signaling : CCS, 22(1), 256-256 (2024-05-06)
Melanoma is a highly heterogeneous cancer, in which frequent changes in activation of signaling pathways lead to a high adaptability to ever changing tumor microenvironments. The elucidation of cancer specific signaling pathways is of great importance, as demonstrated by the
Elisa Guffanti et al.
Journal of cell science, 121(Pt 12), 2037-2045 (2008-05-29)
Nucleolar channel systems (NCSs) are membranous organelles appearing transiently in the epithelial cell nuclei of postovulatory human endometrium. Their characterization and use as markers for a healthy receptive endometrium have been limited because they are only identifiable by electron microscopy.
Jun Yang et al.
Nature microbiology, 2, 17086-17086 (2017-06-06)
Chronic oxidative injury produced by airway disease triggers a transforming growth factor-β (TGF-β)-mediated epigenetic reprogramming known as the epithelial-mesenchymal transition (EMT). We observe that EMT silences protective mucosal interferon (IFN)-I and III production associated with enhanced rhinovirus (RV) and respiratory
Shannon E Weigum et al.
BMC neuroscience, 4, 6-6 (2003-04-17)
Nuclear objects that have in common the property of being recognized by monoclonal antibodies specific for phosphoprotein epitopes and cytoplasmic intermediate filaments (in particular, SMI-31 and RT-97) have been reported in glial and neuronal cells, in situ and in vitro.
Oliver Dreesen et al.
The Journal of cell biology, 200(5), 605-617 (2013-02-27)
The nuclear lamina consists of A- and B-type lamins. Mutations in LMNA cause many human diseases, including progeria, a premature aging syndrome, whereas LMNB1 duplication causes adult-onset autosomal dominant leukodystrophy (ADLD). LMNB1 is reduced in cells from progeria patients, but

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service