Skip to Content
Merck
All Photos(1)

Key Documents

MAB3402X

Sigma-Aldrich

Anti-Glial Fibrillary Acidic Protein Antibody, clone GA5, Alexa Fluor 488

clone GA5, Chemicon®, from mouse

Synonym(s):

GFAP

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

mouse

Quality Level

conjugate

ALEXA FLUOR 488

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

GA5, monoclonal

species reactivity

human, chicken, pig, rat

manufacturer/tradename

Chemicon®

technique(s)

immunocytochemistry: suitable
immunohistochemistry: suitable

isotype

IgG1

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... GFAP(2670)

Specificity

The antibody reacts with GFAP from human, pig, chicken and rat. In tissue sections this antibody stains astrocytes and Bergman glia cells (1).

Immunogen

Purified glial filament (1).

Application

Detect Glial Fibrillary Acidic Protein using this Anti-Glial Fibrillary Acidic Protein Antibody, clone GA5, Alexa Fluor 488 validated for use in IC, IH.
Immunocytochemistry

Immunohistochemistry

Optimal working dilutions must be determined by end user.
Research Category
Neuroscience
Research Sub Category
Neuronal & Glial Markers

Physical form

Purified immunoglobulin conjugated to Alexa Fluor 488. Liquid in Phosphate buffer with 15 mg/mL BSA as a stabilizer and 0.1% sodium azide.

Storage and Stability

Maintain at 2-8°C in the dark for up to 6 months after date of receipt.

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Legal Information

ALEXA FLUOR is a trademark of Life Technologies
CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Alexa Fluor is a registered trademark of Molecular Probes, Inc.

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Francesco De Logu et al.
BMC biology, 18(1), 197-197 (2020-12-16)
The mechanism underlying the pain symptoms associated with chemotherapeutic-induced peripheral neuropathy (CIPN) is poorly understood. Transient receptor potential ankyrin 1 (TRPA1), TRP vanilloid 4 (TRPV4), TRPV1, and oxidative stress have been implicated in several rodent models of CIPN-evoked allodynia. Thalidomide
Anna G McNally et al.
Frontiers in molecular neuroscience, 9, 11-11 (2016-02-24)
The consolidation of short-term labile memories for long-term storage requires transcription and there is growing interest in defining the epigenetic mechanisms regulating these transcriptional events. In particular, it has been hypothesized that combinations of histone post-translational modifications (PTMs) have the
Chongyang Ma et al.
Frontiers in pharmacology, 11, 519-519 (2020-05-28)
Stroke is the second leading cause of death after heart disease globally and cerebral ischemic stroke accounts for approximately 70% of all incident stroke cases. We selected four main compounds from a patent Chinese medicine, Qingkailing (QKL) injection, including baicalin
Yang Xu et al.
Scientific reports, 6, 27512-27512 (2016-06-11)
Patients with tumors that metastasize to bone frequently suffer from debilitating pain, and effective therapies for treating bone cancer are lacking. This study employed a novel strategy in which herpes simplex virus (HSV) carrying a small interfering RNA (siRNA) targeting
Kerry-Ann A Stewart et al.
Journal of neuropathology and experimental neurology, 69(12), 1210-1219 (2010-11-26)
Viral infection of the central nervous system can lead to long-term neurologic defects, including increased risk for the development of epilepsy. We describe the development of the first mouse model of viral-induced epilepsy after intracerebral infection with Theiler's murine encephalomyelitis

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service