Skip to Content
Merck
All Photos(1)

Key Documents

MAB2079Z

Sigma-Aldrich

Anti-Integrin β1 Antibody, activated, clone HUTS-4, Azide Free

clone HUTS-4, Chemicon®, from mouse

Synonym(s):

Ligand-inducedBindingSite, LIBS, CLIBS, MAB2079

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

mouse

Quality Level

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

HUTS-4, monoclonal

species reactivity

rat, human

manufacturer/tradename

Chemicon®

technique(s)

ELISA: suitable
flow cytometry: suitable
immunohistochemistry: suitable
western blot: suitable

isotype

IgG2b

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... ITGB1(3688)
rat ... Itgb1(24511)

General description

Integrin beta 1, also known as CD29, is a 130 kDa transmembrane glycoprotein that forms noncovalent complexes with various Integrin alpha subunits (including alpha 1, alpha 2, alpha 3, alpha 4, alpha 5, and alpha 6) to form the functional receptors that bind to specific extracellular matrix proteins. Integrin receptors are involved in the regulation of a variety of important biological functions, including embryonic development, wound repair, hemostasis, and prevention of programmed cell death. They are also implicated in abnormal pathological states such as tumor directed angiogenesis, tumor cell growth, and metastasis. These heterodimeric receptors bridge the cytoplasmic actin cytoskeleton with proteins present in the extracellular matrix and/or on adjacent cells. Interactions between integrins and the extracellular matrix lead to activation of signal transduction pathways and regulation of gene expression.

Specificity

Monoclonal antibody MAB2079Z is specific for active conformation of human beta1 integrins. The beta1 or VLA integrin subgroup contains at least 10 members, each with a distinct alpha subunit nonconvalently associated with the common beta1 subunit. Functional activation of integrins can be experimentally induced by addition of Mg2+ or Mn2+, or by using one of several stimulatory antibodies that are available. Intracellular signals generated as a result of cell activation also modulate the confirmation and affinity of integrins in cellular systems. Antibodies such as MAB2079Z which have the ability to discriminate between different states of integrin activation are exceptionally useful for studies on the regulation of activation. Antibody MAB2079Z recognizes epitopes in the 355-425 region of the common beta1 subunit of VLA integrins, whose expression parallels the ligand binding activity of these adhesion receptors induced by various extracellular and intracellular stimuli.

Immunogen

Epitope: activated, azide free
Generated against human beta1 integrins purified from Triton X-100 lysates of human lung, liver, and skeletal muscle tissues. MAB2079Z was specifically selected due to its increased reactivity with T-lymphoblasts upon Mn2+ stimulation.

Application

Detect Integrin β1 using this Anti-Integrin β1 Antibody, activated, clone HUTS-4, Azide Free validated for use in ELISA, FC, IH and WB.
ELISA

Immunohistochemistry: fresh frozen, or light fixation, with 4% PFA permeabilized with 0.1% triton X-100 in the presence of Mn++ as the epitope of HUTS-4 is dependent upon Mn++ ion. Note antibody only stains activated beta1 integrins. Activated beta-1 integrins are typically found in wound regions for instance (see http://www.ncbi.nlm.nih.gov/books/bv.fcgi?rid=eurekah.figgrp.12951) . Use MAB1981 or CBL1333 for general beta1 integrin staining and detection.

Western Blotting: non-reducing conditions only (see: http://www.jbc.org/cgi/content/full/271/19/11067).

FACS analysis: 2.5 μg antibody / 5 x 105 cells.
Research Category
Cell Structure
Research Sub Category
Integrins

Target description

88 kDa

Physical form

Format: Purified
Protein A Purified mouse immunoglobulin in PBS containing no preservatives.
Protein A purified

Storage and Stability

Maintain for 1 year at 2–8°C from date of shipment. For maximum recovery of product, centrifuge the original vial after thawing and prior to removing the cap.

Analysis Note

Control
Human lung, liver, and skeletal muscle tissues

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Legal Information

CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Bo Wu et al.
Journal of experimental & clinical cancer research : CR, 34, 110-110 (2015-10-07)
Dysregulated endocytosis of membrane proteins contributes significantly to several hallmarks of cancer. Basigin can enhance cancer progression, but its precise mechanism remains unclear. CD98 promotes cell spreading and tumorigenicity by triggering integrin clustering and enhancing cell adhesion to the extracellular
Honghong Yao et al.
FASEB journal : official publication of the Federation of American Societies for Experimental Biology, 27(4), 1532-1548 (2013-01-08)
One of the hallmark features of HIV-associated neurological disease is increased activation and migration of microglia. HIV transactivator of transcription (Tat) is released from infected cells and has the ability to recruit microglia. The purpose of this study was to
Li-Yan Li et al.
Oncotarget, 6(18), 15940-15952 (2015-04-25)
The paucity of new drugs for the treatment of esophageal squamous cell carcinoma (ESCC) limits the treatment options. This study characterized the therapeutic efficacy and action mechanism of a novel natural macrolide compound F806 in human ESCC xenograft models and
Xiaoyu Xiang et al.
NPJ precision oncology, 2(1), 2-2 (2018-06-07)
Extracellular matrix (ECM)-induced β1-integrin-FAK signaling promotes cell attachment, survival, and migration of cancer cells in a distant organ so as to enable cancer metastasis. However, mechanisms governing activation of the β1-integrin-FAK signaling remain incompletely understood. Here, we report that vasodilator-stimulated
Congcong Shen et al.
International journal of molecular medicine, 39(4), 949-959 (2017-03-16)
The commercial use of epidermal growth factor (EGF) is extensive and has been shown to be effective for skin wound healing in clinical practice. There is evidence to indicate that the topical administration of EGF significantly accelerates re-epithelialization by promoting keratinocyte

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service