Skip to Content
Merck
All Photos(1)

Key Documents

MAB2075Z

Sigma-Aldrich

Anti-Integrin β6 Antibody, clone R6G9, azide free

clone R6G9, Chemicon®, from mouse

Synonym(s):

MAB2075

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

mouse

Quality Level

antibody form

purified antibody

antibody product type

primary antibodies

clone

R6G9, monoclonal

species reactivity

human

manufacturer/tradename

Chemicon®

technique(s)

flow cytometry: suitable
immunoprecipitation (IP): suitable

isotype

IgG2a

suitability

not suitable for Western blot
not suitable for activity/function inhibition

NCBI accession no.

UniProt accession no.

shipped in

dry ice

target post-translational modification

unmodified

Gene Information

human ... ITGB6(3694)

Specificity

Monoclonal antibody MAB2075Z, raised by immunizing Balb/c mice with murine NIH 3T3 cells stably transfected with the human beta6 subunit, recognizes the human beta6 subunit by flow cytometry and is an excellent antibody for immunoprecipitation.

Immunogen

Murine NIH 3T3 cells stably transfected with the human (6 subunit

Application

Anti-Integrin β6 Antibody, clone R6G9, azide free is an antibody against Integrin β6 for use in FC & IP.
Flow cytometry

Immunoprecipitation: Effective; however because this antibody is an IgG2a, protein G or rabbit anti-mouse IgG is helpful for capture.

Immunohistochemistry/immunocytochemistry: The antibody recognizes alphaVbeta6 by immunohistochemistry and immunocytochemistry (acetone fix), but nuclear background staining limits usefulness in these techniques.


Then antibody is not function blocking and not effective for western blot.

Optimal working dilutions must be determined by end user.

Physical form

Format: Purified

Legal Information

CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Yasuyuki Taooka et al.
Respiration; international review of thoracic diseases, 86(5), 367-375 (2012-12-22)
Repeated aspiration pneumonia is a serious problem in the elderly. In aspiration pneumonia, neutrophils play an important role in acute lung injury, while CD18-independent neutrophil transmigration pathways have also been reported in acid-aspiration pneumonia animal models. However, the involvement of
Alison Burman et al.
Journal of virology, 80(19), 9798-9810 (2006-09-16)
Foot-and-mouth disease virus (FMDV) can use a number of integrins as receptors to initiate infection. Attachment to the integrin is mediated by a highly conserved arginine-glycine-aspartic acid (RGD) tripeptide located on the GH loop of VP1. Other residues of this
Terry Jackson et al.
Journal of virology, 78(9), 4533-4540 (2004-04-14)
Field isolates of foot-and-mouth disease virus (FMDV) have been shown to use three alphav integrins, alphavbeta1, alphavbeta3, and alphavbeta6, as cellular receptors. Binding to the integrin is mediated by a highly conserved RGD motif located on a surface-exposed loop of
Liudmila S Chesnokova et al.
Proceedings of the National Academy of Sciences of the United States of America, 106(48), 20464-20469 (2009-11-19)
Epstein-Barr virus (EBV) is a ubiquitous human herpesvirus that is causally implicated in the development of lymphoid and epithelial tumors. Entry of virus requires fusion of virus envelopes and cell membranes. Fusion with B lymphocytes requires virus glycoprotein gB and
John J Grzesiak et al.
International journal of cancer, 129(12), 2905-2915 (2011-04-15)
To address the role of β(1) integrins in pancreatic cancer progression, we stably knocked down β(1) integrin subunit expression in human FG-RFP pancreatic cancer cells using lentiviral-based RNA interference. We then examined the effects of β(1) integrin

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service