Skip to Content
Merck
All Photos(1)

Key Documents

MAB1953

Sigma-Aldrich

Anti-Integrin αV Antibody, clone P3G8

clone P3G8, Chemicon®, from mouse

Synonym(s):

CD51

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

mouse

Quality Level

antibody form

purified antibody

antibody product type

primary antibodies

clone

P3G8, monoclonal

species reactivity

human

manufacturer/tradename

Chemicon®

technique(s)

ELISA: suitable
flow cytometry: suitable
immunocytochemistry: suitable
immunohistochemistry: suitable
immunoprecipitation (IP): suitable

isotype

IgG1

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... ITGAV(3685)

Specificity

Reacts with human alphaV integrin subunit and with all alphaV-containing integrin receptors.

Immunogen

UCLA P3 lung carcinoma cells

Application

Anti-Integrin αV Antibody, clone P3G8 is an antibody against Integrin αV for use in ELISA, FC, IP, IC, IH.
Immunohistochemistry: 1:1,000. For use on acetone or paraformaldehyde fixed tissue

Immunocytochemistry: 1:1,000. Will react with some lymphoid cell lines (B cells), many carcinoma and melanoma cell lines and osteosarcomas.

Flow cytometry

Immunoprecipitation

FACS

EIA

Optimal working dilutions must be determined by end user.
Research Category
Cell Structure
Research Sub Category
Integrins

Physical form

Format: Purified

Storage and Stability

Maintain at 2-8°C in undiluted aliquots for up to 6 months.

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Legal Information

CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Maria Rita Milone et al.
Oncotarget, 6(7), 5324-5341 (2014-12-08)
Proteomic analysis identified differentially expressed proteins between zoledronic acid-resistant and aggressive DU145R80 prostate cancer (PCa) cells and their parental DU145 cells. Ingenuity Pathway Analysis (IPA) showed a strong relationship between the identified proteins within a network associated with cancer and
Keijiro Ishikawa et al.
FASEB journal : official publication of the Federation of American Societies for Experimental Biology, 28(1), 131-142 (2013-09-12)
Proliferative vitreoretinopathy (PVR) is a severe, vision-threatening disorder characterized by the fibrous membrane formation that leads to tractional retinal detachment. There has been no effective therapeutic approach other than vitreoretinal surgery. In this study, DNA microarray analysis of the fibrous
D H Chai et al.
Osteoarthritis and cartilage, 18(2), 249-256 (2009-10-06)
Our goal was to test the hypothesis that specific integrin receptors regulate chondrocyte biosynthetic response to dynamic compression at early times in 3D gel culture, during initial evolution of the pericellular matrix, but prior to significant accumulation of further-removed matrix.
Mannose 6-phosphate/insulin-like growth factor 2 receptor limits cell invasion by controlling alphaVbeta3 integrin expression and proteolytic processing of urokinase-type plasminogen activator receptor.
Schiller, HB; Szekeres, A; Binder, BR; Stockinger, H; Leksa, V
Molecular Biology of the Cell null
Tropism-modified AAV vectors overcome barriers to successful cutaneous therapy.
Sallach, J; Di Pasquale, G; Larcher, F; Niehoff, N; Rubsam, M; Huber, A; Chiorini et al.
Molecular Therapy null

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service