Skip to Content
Merck
All Photos(1)

Key Documents

MAB1615

Sigma-Aldrich

Anti-Neurofilament 70 kDa Antibody, clone DA2

culture supernatant, clone DA2, Chemicon®

Synonym(s):

Anti-CMT1F, Anti-CMTDIG, Anti-NF-L, Anti-NF68, Anti-NFL, Anti-PPP1R110

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

mouse

Quality Level

antibody form

culture supernatant

antibody product type

primary antibodies

clone

DA2, monoclonal

species reactivity

mouse, bovine, rat, human, pig

manufacturer/tradename

Chemicon®

technique(s)

immunocytochemistry: suitable
immunohistochemistry: suitable
western blot: suitable

isotype

IgG1

NCBI accession no.

UniProt accession no.

shipped in

dry ice

target post-translational modification

unmodified

Gene Information

human ... NEFL(4747)

General description

Neurofilaments are a type of intermediate filament that serve as major elements of the cytoskeleton supporting the axon cytoplasm. They are the most abundant fibrillar components of the axon, being on average 3-10 times more frequent than axonal microtubules. Neurofilaments (10nm in dia.) are built from three intertwined protofibrils which are themselves composed of two tetrameric protofilament complexs of monomeric proteins. The neurofilament triplet proteins (68/70, 160, and 200 kDa) occur in both the central and peripheral nervous system and are usually neuron specific. The 68/70 kDa NF-L protein can self-assemble into a filamentous structure, however the 160 kDa NF-M and 200 kDa NF-H proteins require the presence of the 68/70 kDa NF-L protein to co-assemble. Neuromas, ganglioneuromas, gangliogliomas, ganglioneuroblastomas and neuroblastomas stain positively for neurofilaments. Although typically restricted to neurons, neurofilaments have been detected in paragangliomas and adrenal and extra-adrenal pheochromocytomas. Carcinoids, neuroendocrine carcinomas of the skin, and oat cell carcinomas of the lung also express neurofilaments. For more neurofilament information see Nervous System Cell Type Specific Marker chart online under the CHEMICON Technical Support section.

Specificity

Recognizes the low molecular weight (68-70 kDa) subunit of the neurofilament triplet (NF-L), the epitope is phosphate independent. Reacts with human and higher vertebrates.

Immunogen

Enzymatically dephosphorylated pig neurofilaments.

Application

Immunoblot and immunohistochemistry (frozen sections, little reactivity on formalin-fixed paraffin embedded material). Optimal working dilutions must be determined by end user.

IMMUNOHISTOCHEMISTRY PROTOCOL FOR MAB1615

This antibody has been used successfully on 30 mm, free floating, 4% paraformaldehyde fixed rat brain tissue. All steps are performed under constant agitation. Suggested protocol follows.

1) 3 x 10 minute washes in TBS (with or without 0.25% Triton).

2) Incubate for 30 minutes in TBS with 3% serum (same as host from secondary antibody).

3) Incubate primary antibody diluted appropriately in TBS with 1% serum (same as host from secondary antibody) (with or without 0.25% Triton) for 2 hours at room temperature followed by 16 hours at 4°C.

4) 3 x 10 minute washes in TBS.

5) Incubate with secondary antibody diluted appropriately in TBS with 1% serum (same as host from secondary antibody).

6) 3 x 10 minute washes in TBS.

7) ABC Elite (1:200 Vector Labs) in TBS.

8) 2 x 10 minute washes in TBS.

9) 1 x 10 minute wash in phosphate buffer (no saline).

10) DAB reaction with 0.06% NiCl added for intensification.

11) 2 x 10 minute washes in PBS.

12) 1 x 10 minute wash in phosphate buffer (no saline).
Research Category
Neuroscience
Research Sub Category
Neurofilament & Neuron Metabolism

Neuronal & Glial Markers
This Anti-Neurofilament 70 kDa Antibody, clone DA2 is validated for use in WB, IC, IH for the detection of Neurofilament 70 kDa.

Linkage

Replaces: 04-1112

Physical form

Liquid. Contains no preservative.

Storage and Stability

Maintain at -20°C in undiluted aliquots for up to 12 months. Avoid repeated freeze-thaw cycles.

Legal Information

CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

I E Kyrou et al.
European journal of vascular and endovascular surgery : the official journal of the European Society for Vascular Surgery, 43(4), 408-414 (2012-01-31)
Heat shock protein 70 (Hsp70) is detected in substantial amounts in normal neurons and this basal content may protect a cell against harmful conditions without the need for additional synthesis. Herein, we investigate the potential protective role of these basal
Renata Novotny et al.
The Journal of neuroscience : the official journal of the Society for Neuroscience, 36(18), 5084-5093 (2016-05-06)
The aggregation of amyloid-β peptide (Aβ) in brain is an early event and hallmark of Alzheimer's disease (AD). We combined the advantages of in vitro and in vivo approaches to study cerebral β-amyloidosis by establishing a long-term hippocampal slice culture
A ROCK Inhibitor Blocks the Inhibitory Effect of Chondroitin Sulfate Proteoglycan on Morphological Changes of Mesenchymal Stromal/Stem Cells into Neuron-Like Cells.
Lim, HS; Joe, YA
Biomolecules & Therapeutics null
Maria Nordheim Alme et al.
Journal of neuroimmunology, 285, 180-186 (2015-07-23)
Fingolimod (FTY720) is approved for treatment of relapsing-remitting multiple sclerosis. In vitro studies have found that fingolimod stimulates remyelination in cerebellar slices, but in vivo animal studies have not detected any positive effect on cerebral remyelination. The discrepant findings could
Catherine M Ivy et al.
The Journal of experimental biology, 223(Pt 5) (2020-02-15)
High-altitude environments are cold and hypoxic, and many high-altitude natives have evolved changes in respiratory physiology that improve O2 uptake in hypoxia as adults. Altricial mammals undergo a dramatic metabolic transition from ectothermy to endothermy in early post-natal life, which

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service