Skip to Content
Merck
All Photos(1)

Key Documents

FCMAB282F

Sigma-Aldrich

Milli-Mark® Anti-Integrin αVβ3 Antibody, clone LM609-FITC

clone LM609, Milli-Mark®, from mouse

Synonym(s):

Vitronectin Receptor, CD51/CD61, integrin alphaVbeta3, alphaVbeta3

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41
Pricing and availability is not currently available.

biological source

mouse

Quality Level

conjugate

FITC conjugate

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

LM609, monoclonal

species reactivity

human

manufacturer/tradename

Milli-Mark®

technique(s)

flow cytometry: suitable

isotype

IgG1

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... ITGAV(3685)

General description

The integrin family of cell adhesion receptors consists of at least 16 membrane-associated heterodimers, composed of an alpha and beta subunit that associate in a non-covalent manner. The structure and functional diversity of the integrin family are based upon the pairing abilities of the individual alpha and beta subunits. Key to these molecular interactions between the integrin receptors and their respective ligands is the recognition of the Arg-Gly-Asp (RGD) sequence, known to be present in the extracellular matrix components fibronectin, vitronectin, collagen, fibrinogen, and von Willebrand factor (Cheresh, 1991). The involvement of integrins in vascular proliferation, adhesion, and wound repair has been well documented. The adhesion receptor, integrin alphaVbeta3, appears to be selectively expressed on growing blood vessels and has been identified as a marker of angiogenic vascular tissue (Brooks, 1994). Due to its involvement in angiogenesis, integrin alphaVbeta3 is one of the most intensely studied of the integrin receptors.

Specificity

Antibody recognizes Integrin αVβ3.

Immunogen

Purified adhesion receptor from the human melanoma cell line M21 (Cherish, D. and Spiro, R. (1987) J. Biol. Chemistry, 262(36):17703-17711).

Application

Detect Integrin αVβ3 using this Milli-Mark Anti-Integrin αVβ3 Antibody, clone LM609-FITC validated for use in FC.
Research Category
Cell Structure
Research Sub Category
Integrins

Quality

Evaluated by flow cytometry using U937 cells.

Physical form

Protein A purified
Purified mouse monoclonal IgG1 conjugated to FITC in PBS with 0.1% sodium azide and 15 mg/mL BSA.

Storage and Stability

Flow Cytometry
Antibody dilution for cellular staining:

• Prepare an antibody working solution by diluting the primary antibody 1:5 with PBS
• Dispense the volume per test of working of solution according to the number of cells indicated in the table below.
• 5 µL for Guava Flow Cytometer
• 10 µL for other Flow Cytometry instruments

Analysis Note

Control
U937 cells

Legal Information

MILLI-MARK is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Katrin Echtler et al.
PloS one, 12(3), e0172788-e0172788 (2017-03-03)
Platelets modulate the process of cancer metastasis. However, current knowledge on the direct interaction of platelets and tumor cells is mostly based on findings obtained in vitro. We addressed the role of the platelet fibrinogen receptor glycoprotein IIb (integrin αIIb)

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service