Skip to Content
Merck
All Photos(1)

Key Documents

EZBRAIN-SET

Millipore

Human Amyloid β40 and Amyloid β42 ELISA Kit

measures Amyloid β40 and Amyloid β42 in 50 μL tissue or cell lysate; combo pack

Synonym(s):

Amyloid β40/β42 ELISA, Brain ELISA Kit, Human Brain Amyloid ELISA

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12161503
NACRES:
NA.84
Pricing and availability is not currently available.

Product Name

Human Amyloid β40 and Amyloid β42 Brain ELISA,

species reactivity

human

Quality Level

packaging

kit of 2 × 96 wells

parameter

50 μL sample volume (Overnight assay)

assay range

sensitivity: 6.0 pg/mL
(The lowest level of Amyloid β1-40 standard used in this assay ; 50 μL sample size)

sensitivity: 8 pg/mL
(The lowest level of Amyloid ß1-42 standard used in this assay; 50 μL sample size)

inter-assay cv: <10
intra-assay cv: <10
standard curve range: 16-500 pg/mL

technique(s)

ELISA: suitable

input

sample type tissue/cellular lysate

application(s)

research use

detection method

colorimetric (450nm/590nm)

shipped in

wet ice

storage temp.

2-8°C

Gene Information

human ... APP(351)

Specificity

The Amyloid β40 ELISA uses monoclonal anti-Aβ antibodies with high selectivity for human Aβ. The capture antibody recognizes the C-terminal end of Amyloid β1-40, which causes a high selectivity for Aβ 40. The cross-reactivity of the used antibodies to other Amyloid peptides was tested by ELISA and BIACORE and shows no significant cross-reactivity to Aβ1-38, Aβ1-39, Aβ1-42, Aβ1-43 and Aβ1-44. The Amyloid β42 ELISA uses monoclonal anti-Aβ antibodies with high selectivity for human Aβ. The capture antibody recognizes the C-terminal end of Amyloid β1-42, which causes a high selectivity for Aβ42. The cross-reactivity of the used antibodies to other Amyloid peptides was tested by ELISA and BIACORE and shows no significant cross-reactivity to Aβ1-38, Aβ1-39, Aβ1-42, Aβ1-43 and Aβ1-44.


Used to detect/quantify: Amyloid β40 & Amyloid β42

Application

These assays require 50 µl of sample and are overnight assays.

Packaging

96-well plate

Storage and Stability

Components in the kit can be stored up to 2 weeks at 2-8°C

Other Notes

Contact technical support for information

Disclaimer

For research use only. Not for use in diagnostic procedures.

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service