Skip to Content
Merck
All Photos(1)

Key Documents

CC12

Sigma-Aldrich

Anti-Cyclin D1 (Ab-3) Mouse mAb (DCS-6)

liquid, clone DCS-6, Calbiochem®

Synonym(s):

Anti-Bcl-1, Anti-PRAD-1

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

mouse

Quality Level

antibody form

purified antibody

antibody product type

primary antibodies

clone

DCS-6, monoclonal

form

liquid

contains

≤0.1% sodium azide as preservative

species reactivity

human, mouse, rat

manufacturer/tradename

Calbiochem®

storage condition

do not freeze

isotype

IgG2a

shipped in

wet ice

storage temp.

2-8°C

target post-translational modification

unmodified

Gene Information

human ... CCND1(595)
mouse ... Ccnd1(12443)
rat ... Ccnd1(58919)

General description

Purified mouse monoclonal antibody generated by immunizing mice with the specified immunogen and fusing splenocytes with NS/1 mouse myeloma cells. Recognizes the cyclin D1 protein.
Recognizes cyclin D1 in MCF-7 and A431 cells. Does not react with cyclins D2 and D3.
This Anti-Cyclin D1 (Ab-3) Mouse mAb (DCS-6) is validated for use in FC, Frozen Sections, Immunoblotting, IF, IP, Neutralization Studies, Paraffin Sections for the detection of Cyclin D1 (Ab-3).

Immunogen

Human
recombinant human cyclin D1

Application

Flow Cytometry (5 µg/ml; Cat. No. CC12, CC12F)

Frozen Sections (see application references)

Immunoblotting (1 µg/ml)

Immunofluorescence (5 µg/ml; Cat. No. CC12, CC12F)

Immunoprecipitation (see application references)

Neutralization Studies (see application references)

Paraffin Sections (5 µg/ml, heat pre-treatment required)

Packaging

Please refer to vial label for lot-specific concentration.

Warning

Toxicity: Standard Handling (A)

Physical form

In 50 mM sodium phosphate buffer, 0.2% gelatin.

Analysis Note

Negative Control
trp E (Ab-1)
Positive Control
MCF7, or A431 cells or breast carcinoma tissue

Other Notes

Does not react with cyclins D2 or D3. Staining of paraffin sections can also be detected with pepsin or trypsin pretreatment; light staining will be detected without pretreatment. Do not microwave sections, as this may destroy staining. For immunofluorescence and flow cytometry, we recommend using the FITC conjugate (Cat. No. CC12F). Antibody should be titrated for optimal results in individual systems.
Inaba, T., et al. 1992. Genomics13, 565.
Jiang, W., et al. 1992. Cancer Res.2, 2980.
Pines, J. 1992. Cell Growth Differ.2, 305.
Xiong, Y., et al. 1992. Genomics13, 575.
Xiong, Y., et al. 1992. Cell71, 504.
Motokura, T., et al. 1991. Nature350, 512.
Solomon, M.J., et al. 1990. Cell63, 1013.
Draetta, G., and Beach, D. 1988. Cell54, 17.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Shumpei Yamada et al.
International journal of cancer, 111(1), 17-22 (2004-06-09)
Cyclin E and Cdk2 have been shown to play an important role in G1/S transition of the cell cycle. Two E-type cyclins (E1 and E2) have been identified to date and share functionally similarities. Upregulation of these cyclins has been
Banumathi K Cole et al.
Molecular and cellular biology, 28(4), 1274-1284 (2007-12-19)
Merlin, the product of the NF2 tumor suppressor gene, is closely related to the ERM (ezrin, radixin, moesin) proteins, which provide anchorage between membrane proteins and the underlying cortical cytoskeleton; all four proteins are members of the band 4.1 superfamily.
Marie-Noëlle Giraud et al.
Journal of applied physiology (Bethesda, Md. : 1985), 99(1), 313-322 (2005-07-23)
Neuregulin/ErbB2-induced kinase signaling provides essential survival and protection clues for functional integrity of the adult heart and skeletal muscle. To define the regulatory pathways involved in neuregulin-dependent muscle cell survival, we set out to map the largely unknown transcript targets
Raanan Berger et al.
Cancer research, 66(11), 5723-5728 (2006-06-03)
The mechanisms underlying the progression of prostate cancer to a state of resistance to hormone ablation remain poorly understood. Here, we have investigated the relationship between androgen receptor (AR) and Her-2/neu in prostate cancer cells. Overexpression of Her-2/neu (c-ErbB2) activates
Yanyuan Wu et al.
Cancer research, 70(13), 5475-5485 (2010-06-17)
Trastuzumab treatment has improved the overall survival of HER2-overexpressing breast cancer patients. However, many of these patients will eventually become resistant to treatment. The mechanisms that contribute to resistance to trastuzumab are unknown. In this study, we tested the hypothesis

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service