Skip to Content
Merck
All Photos(1)

Key Documents

CA1023

Sigma-Aldrich

Anti-Hsp90α Mouse mAb (EMD-17D7)

liquid, clone EMD-17D7, Calbiochem®

Synonym(s):

Anti-Heat Shock Protein 90α

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

mouse

Quality Level

antibody form

purified antibody

antibody product type

primary antibodies

clone

EMD-17D7, monoclonal

form

liquid

contains

≤0.1% sodium azide as preservative

species reactivity

human

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
avoid repeated freeze/thaw cycles

isotype

IgG1

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

General description

Protein G PLUS/protein A purified mouse monoclonal antibody. Recognizes the ~90 kDa Hsp90α protein.
Recognizes the ~90 kDa Hsp90α protein in HeLa cells. Does not recognize Hsp90β.
This Anti-Hsp90α Mouse mAb (EMD-17D7) is validated for use in ELISA, Immunoblotting, Immunoprecipitation for the detection of Hsp90α.

Immunogen

Hsp90α, His•Tag, Human, Recombinant (Cat. No. 385901)
Human

Application



ELISA (see comments)
Immunoblotting (1 g/ml)
Immunoprecipitation (5 g/0.7 mg protein)

Packaging

Please refer to vial label for lot-specific concentration.

Warning

Toxicity: Irritant (B)

Physical form

In PBS.

Reconstitution

Following initial thaw, aliquot and freeze (-20°C).

Analysis Note

Positive Control
H460 and HeLa cells

Other Notes

Detects native and recombinant Hsp90α. Does not recognize Hsp90β. This antibody will also work for ELISA, but concentration is assay dependent. Antibody should be titrated for optimal results in individual systems.
Scheibel, T., et al. 1999. Proc. Natl. Acad. Sci. USA96, 1297.
Nemoto, T., et al. 1997 J. Biol. Chem.272, 26179.
Nemoto, T., et al. 1998. Biochem. J.330, 989.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Wei Li et al.
The EMBO journal, 26(5), 1221-1233 (2007-02-17)
Hypoxia is a microenvironmental stress in wounded skin, where it supports wound healing by promoting cell motility. The mechanism of the hypoxia action remained speculative. Here, we provide evidence that hypoxia promotes human dermal fibroblast (HDF) migration by inducing secretion
Lin Liu et al.
Cancer science, 114(10), 4020-4031 (2023-08-23)
Lipids are a major component of extracellular vesicles; however, their significance in tumorigenesis and progression has not been well elucidated. As we previously found that lipid profiles drastically changed in breast tumors upon progression, we hypothesized that lipid profiles of
Priyamvada Jayaprakash et al.
Journal of cell science, 128(8), 1475-1480 (2015-03-05)
When tissues are injured and blood vessels clot, the local environment becomes ischemic, meaning that there is a lack of adequate supply of oxygen and glucose delivered to the surrounding cells. The heat shock protein-90 (Hsp90) family proteins protect tissues
Hang-Ming Dong et al.
Respiratory research, 18(1), 111-111 (2017-06-01)
The disruption and hyperpermeability of bronchial epithelial barrier are closely related to the pathogenesis of asthma. House dust mite (HDM), one of the most important allergens, could increase the airway epithelial permeability. Heat shock protein (Hsp) 90α is also implicated
Julian D Gillmore et al.
Journal of the American Society of Nephrology : JASN, 20(2), 444-451 (2008-12-17)
Mutations in the fibrinogen A alpha-chain gene are the most common cause of hereditary renal amyloidosis in the United Kingdom. Previous reports of fibrinogen A alpha-chain amyloidosis have been in isolated kindreds, usually in the context of a novel amyloidogenic

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service