Skip to Content
Merck
All Photos(1)

Key Documents

AX0078

Supelco

Acetic acid

OmniTrace Ultra, 99%, for inorganic trace analysis

Synonym(s):

aa, Glacial acetic acid

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3CO2H
CAS Number:
Molecular Weight:
60.05
Beilstein:
506007
MDL number:
UNSPSC Code:
12352106
EC Index Number:
200-580-7
Pricing and availability is not currently available.

grade

for inorganic trace analysis

Quality Level

vapor density

2.07 (vs air)

vapor pressure

15.4 hPa ( 20 °C)

Assay

99%

form

liquid

autoignition temp.

800 °F

expl. lim.

16 %, 92 °F
4 %, 59 °F

refractive index

n20/D 1.371 (lit.)

pH

2.5 (20 °C, 50 g/L in H2O)

bp

117-118 °C (lit.)

mp

16.2 °C (lit.)

transition temp

flash point 39 °C

solubility

602.9 g/L

density

1.04 g/mL at 25 °C (lit.)

cation traces

Ag: ≤50 pg/g
Al: ≤50 pg/g
As: ≤50 pg/g
Ba: ≤10 pg/g
Be: ≤10 pg/g
Bi: ≤10 pg/g
Ca: ≤50 pg/g
Cd: ≤10 pg/g
Ce: ≤10 pg/g
Co: ≤10 pg/g
Cr: ≤10 pg/g
Cs: ≤10 pg/g
Cu: ≤10 pg/g
Dy: ≤1 pg/g
Er: ≤1 pg/g
Eu: ≤1 pg/g
Fe: ≤50 pg/g
Ga: ≤10 pg/g
Gd: ≤1 pg/g
Ge: ≤10 pg/g
Hf: ≤10 pg/g
Ho: ≤1 pg/g
In: ≤1 pg/g
K: ≤50 pg/g
La: ≤1 pg/g
Li: ≤10 pg/g
Lu: ≤10 pg/g
Mg: ≤50 pg/g
Mn: ≤10 pg/g
Mo: ≤10 pg/g
Na: ≤100 pg/g
Nd: ≤1 pg/g
Ni: ≤50 pg/g
Pb: ≤10 pg/g
Pr: ≤1 pg/g
Pt: ≤50 pg/g
Rb: ≤10 pg/g
Re: ≤10 pg/g
Rh: ≤50 pg/g
Ru: ≤50 pg/g
Sb: ≤50 pg/g
Sc: ≤10 pg/g
Sm: ≤1 pg/g
Sn: ≤50 pg/g
Sr: ≤10 pg/g
Tb: ≤1 pg/g
Te: ≤1 pg/g
Th: ≤1 pg/g
Ti: ≤10 pg/g
Tl: ≤10 pg/g
Tm: ≤1 pg/g
U: ≤1 pg/g
V: ≤10 pg/g
W: ≤10 pg/g
Y: ≤1 pg/g
Yb: ≤1 pg/g
Zn: ≤50 pg/g
Zr: ≤10 pg/g

SMILES string

[F2C(F2C)13F3C]C(O)=O

shipped in

ambient

storage temp.

room temp

InChI

1S/C2H4O2/c1-2(3)4/h1H3,(H,3,4)

InChI key

QTBSBXVTEAMEQO-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

Application

OmniTrace Ultra acids for trace metal analysis and sample preparation. These acids are tested to PPT (parts per trillion) levels.

Packaging

Product is double-distilled and packed in class 100 clean room conditions into preleached FEP bottles. Each FEP bottle is packed in an EPS innerpack and then packed into a 1x regulated carton with a Certificate of Analysis.

Pictograms

FlameCorrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Flam. Liq. 3 - Skin Corr. 1A

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

102.2 °F - closed cup

Flash Point(C)

39 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service