Skip to Content
Merck
All Photos(1)

Documents

AP181R

Millipore

Goat Anti-Mouse IgG Antibody, Rhodamine conjugate, Species Adsorbed

1.4 mg/mL, Chemicon®

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.46

biological source

goat

Quality Level

conjugate

TRITC conjugate
rhodamine conjugate

antibody form

affinity isolated antibody

antibody product type

secondary antibodies

clone

polyclonal

species reactivity

mouse

manufacturer/tradename

Chemicon®

concentration

1.4 mg/mL

technique(s)

immunofluorescence: suitable

shipped in

wet ice

target post-translational modification

unmodified

Specificity

Mouse IgG (H+L) (absorbed for minimal cross-reactivity to Rat, Human, Bovine and Horse Serum Proteins)

FLUOROPHORE/ PROTEIN ABSORBANCE RATIO:

Tetramethyl Rhodamine Isothiocyanate A550/A280 = 0.57

Application

Goat anti-Mouse IgG Antibody, Rhodamine conjugate, Species Adsorbed detects level of Mouse IgG & has been published & validated for use in IF.

Physical form

Lyophilized. Buffer = 0.01 M Sodium Phosphate, 0.25 M NaCl, pH 7.6 and 0.05%Sodium azide.

RECONSTITUTION:

Reconstitute to 1 mg/mL with sterile distilled water.

Legal Information

CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany

Not finding the right product?  

Try our Product Selector Tool.

Hazard Statements

Precautionary Statements

Hazard Classifications

Aquatic Chronic 3

Storage Class Code

11 - Combustible Solids

WGK

WGK 3


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Danielle A Guimarães et al.
Redox biology, 6, 386-395 (2015-09-08)
Imbalanced matrix metalloproteinase (MMP)-2 activity and transforming growth factor expression (TGF-β) are involved in vascular remodeling of hypertension. Atorvastatin and sildenafil exert antioxidant and pleiotropic effects that may result in cardiovascular protection. We hypothesized that atorvastatin and sildenafil alone or
Yuan Li et al.
Molecular medicine reports, 15(5), 2673-2680 (2017-04-28)
Advanced glycation end products (AGEs) restrain the proliferation of endothelial cells, which is an important determinant of diabetic vasculopathy. Mitochondrial biogenesis serves an essential role in cellular adaptation and repair. The current study aimed to investigate alterations in mitochondrial energy
Natsumi Maruyama et al.
The Journal of reproduction and development, 69(6), 328-336 (2023-11-06)
Senescent cells play a detrimental role in age-associated pathogenesis by producing factors involved in senescence-associated secretory phenotype (SASP). The present study was conducted to examine the possibility that senescent cells are present in aged ovaries and, if so, to determine
Ye Chang et al.
International journal of molecular medicine, 41(1), 33-42 (2017-11-09)
This study aimed to explore whether angiotensin II (Ang II) inhibits the proliferation of human umbilical vein endothelial cells (HUVECs) by changing mitochondrial energy metabolism, and whether atorvastatin has a protective role via restoration of endothelial function. HUVECs were treated with 1 µM Ang II alone
Selim Cellek et al.
Gastroenterology, 133(1), 175-183 (2007-07-17)
Although the beta(3)-adrenoceptor (AR) has been suggested to be involved in regulation of gut motility and visceral algesia, the precise mechanisms have been unknown. beta(3)-AR has been postulated to have a nonneuronal expression, being initially characterized in adipocytes and subsequently

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service