Skip to Content
Merck
All Photos(1)

Key Documents

AG208

Sigma-Aldrich

VGT 1 Peptide

control for AB5905, liquid

Synonym(s):

VGLUT1, BNPI

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.42

product name

Vesicular Glutamate Transporter 1, control peptide for AB5905,

biological source

rat

Quality Level

form

liquid

manufacturer/tradename

Chemicon®

concentration

1 mg/mL

technique(s)

cell based assay: suitable

NCBI accession no.

UniProt accession no.

shipped in

dry ice

General description

Synthetic peptide from rat VGLUT1 protein.
VGLUT1 is expressed in a subset of glutamate neurons and transports glutamate into native synaptic vesicles from the brain, exhibiting a conductance for chloride that is blocked by glutamate. Vesicular glutamate transport has a substantially lower apparent affinity than the plasma membrane excitatory amino acid transporters. Glutamate transport by VGLUT1 is saturated with a K(m) of approximately 2 mM, in the same range as transport by synaptic vesicles. Finally, plasma membrane glutamate transporters recognize both aspartate and glutamate as substrates, whereas VGLUT1 does not recognize aspartate.

Application

For use in blocking the reactivity of AB5905.

Optimal working dilution must be determined by the end user.

Legal Information

CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

12 - Non Combustible Liquids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Sean P Masterson et al.
The Journal of comparative neurology, 515(6), 647-663 (2009-06-06)
Dorsal thalamic nuclei have been categorized as either "first-order" nuclei that gate the transfer of relatively unaltered signals from the periphery to the cortex or "higher order" nuclei that transfer signals from one cortical area to another. To classify the
Ramon Guirado et al.
Heliyon, 4(6), e00669-e00669 (2018-07-14)
The quantification of the expression of different molecules is a key question in both basic and applied sciences. While protein quantification through molecular techniques leads to the loss of spatial information and resolution, immunohistochemistry is usually associated with time-consuming image
Seth L Shipman et al.
Proceedings of the National Academy of Sciences of the United States of America, 109(47), 19432-19437 (2012-11-07)
The transsynaptic complex of neuroligin (NLGN) and neurexin forms a physical connection between pre- and postsynaptic neurons that occurs early in the course of new synapse assembly. Both neuroligin and neurexin have, indeed, been proposed to exhibit active, instructive roles
VGLUT1 and VGLUT2 innervation in autonomic regions of intact and transected rat spinal cord.
Ida J Llewellyn-Smith,Carolyn L Martin,Natalie M Fenwick,Stephen E Dicarlo,Heidi L Lujan et al.
The Journal of Comparative Neurology null
Synaptotagmins I and II in the developing rat auditory brainstem: Synaptotagmin I is transiently expressed in glutamate-releasing immature inhibitory terminals.
Alan P Cooper,Deda C Gillespie
The Journal of Comparative Neurology null

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service