Skip to Content
Merck
All Photos(1)

Key Documents

ABT146

Sigma-Aldrich

Anti-Occludin Antibody

from rabbit, purified by affinity chromatography

Synonym(s):

Occludin

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

rabbit

Quality Level

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

purified by

affinity chromatography

species reactivity

human

species reactivity (predicted by homology)

bovine (based on 100% sequence homology), mouse (based on 100% sequence homology), porcine (based on 100% sequence homology), canine (based on 100% sequence homology), primate (based on 100% sequence homology), rat (based on 100% sequence homology), equine (based on 100% sequence homology)

packaging

antibody small pack of 25 μg

technique(s)

western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

ambient

target post-translational modification

unmodified

Gene Information

General description

Occludin is a 65-kDa integral plasma-membrane protein located at the tight junctions. It is required for cytokine-induced regulation of the tight junction paracellular permeability barrier. Occludin is believed to prevent the transport of macromolecules through the tight junction. Encoded by the OLCN gene, occludin spans the plasma membrane four times, forming two extracellular loops and exposing its NH2 and COOH terminus to the cytosol. This cytoplasmic tail binds directly to the ZO family of proteins, which link the protein complex to the actin cytoskeleton. Phosphorylation/dephosphorylation plays a major role in regulation of occludin and tight junctions. Expression of occludin is required for infection of liver cells by hepatitis C virus (HCV).

Specificity

This antibody recognizes the N-terminus of Occludin.

Immunogen

Epitope: N-terminus
KLH-conjugated linear peptide corresponding to the N-terminus of human Occludin.

Application

Research Category
Cell Structure
Research Sub Category
ECM Proteins
This Anti-Occludin Antibody is validated for use in Western Blotting for the detection of Occludin.

Quality

Evaluated by Western Blot in human kidney tissue lysate.

Western Blot Analysis: 0.2 µg/mL of this antibody detected Occludin in 10 µg of human kidney tissue lysate.

Target description

~63 kDa observed. The calculated molecular weight is 63 kDa, however Occludin has been shown ranging from ~55 to ~65 kDa in western blots (Florian, P., et al. (2003). Biochemical and Biophysical Research Communications. 304:5-10).

Physical form

Affinity purified
Purified rabbit polyclonal in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Analysis Note

Control
Human kidney tissue lysate

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Duobin Zhang et al.
Journal of leukocyte biology, 111(5), 953-966 (2021-09-30)
Ischemic stroke (IS) can cause disability and death, and microglia as the immune component of the CNS can release inflammatory factors and participate in blood-brain barrier (BBB) dysfunction. This study aimed to investigate the effects of long noncoding RNA (lncRNA)
Hengameh Chloé Mirsepasi-Lauridsen et al.
Clinical and translational gastroenterology, 7, e149-e149 (2016-03-05)
The potential of Escherichia coli (E. coli) isolated from inflammatory bowel disease (IBD) patients to damage the integrity of the intestinal epithelium was investigated. E. coli strains isolated from patients with ulcerative colitis (UC) and healthy controls were tested for
Amy L Creekmore et al.
Pain, 159(9), 1777-1789 (2018-06-19)
In humans, chronic psychological stress is associated with increased intestinal paracellular permeability and visceral hyperalgesia, which is recapitulated in the chronic intermittent water avoidance stress (WAS) rat model. However, it is unknown whether enhanced visceral pain and permeability are intrinsically
Maj Schneider Thomsen et al.
PloS one, 16(3), e0236770-e0236770 (2021-03-13)
The blood-brain barrier (BBB) is formed by brain capillary endothelial cells (BECs) supported by pericytes and astrocytes. The BBB maintains homeostasis and protects the brain against toxic substances circulating in the blood, meaning that only a few drugs can pass
Diana Duarte Lobo et al.
Acta neuropathologica communications, 8(1), 152-152 (2020-09-02)
Blood-brain barrier (BBB) disruption is a common feature in neurodegenerative diseases. However, BBB integrity has not been assessed in spinocerebellar ataxias (SCAs) such as Machado-Joseph disease/SCA type 3 (MJD/SCA3), a genetic disorder, triggered by polyglutamine-expanded ataxin-3. To investigate that, BBB

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service