Skip to Content
Merck
All Photos(1)

Key Documents

ABN100

Sigma-Aldrich

Anti-Vesicular Acetylcholine Transporter (VAChT) Antibody

from goat, purified by affinity chromatography

Synonym(s):

Vesicular acetylcholine transporter, VAChT, rVAT, Solute carrier family 18 member 3

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41
Pricing and availability is not currently available.

biological source

goat

Quality Level

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

purified by

affinity chromatography

species reactivity

mouse, rat

technique(s)

ELISA: suitable
immunohistochemistry: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

Related Categories

General description

Acetylcholine mediates cholinergic transmission in the central and peripheral nervous systems, and is an important modulator of cognition, movement, and cardiovascular function, among other processes. The effects of acetylcholine are regulated by its synthesis, storage, and breakdown, which involve the related enzymes choline acetyltransferase (ChAT), vesicular acetylcholine transporter (VAChT), and acetylcholinesterase (AChE), respectively. VAChT is primarily found in the membrane of presynaptic secretory vesicles. It contains 12 transmembrane domains and a synaptic-vesicular–targeting signal located on the intracellular C-terminal tail. The primary function of VAChT is to actively transport cytoplasmic ACh, across an electrochemical gradient, into synaptic vesicles that release ACh upon cell stimulation. The VAChT can affect cholinergic signaling by affecting the relative pools of ACh secreted to activate postsynaptic nicotinic and muscarinic receptors. VAChT is phosphorylated on its serine 480 residue in the C-terminal tail by protein kinase C. Although the precise role of this modification is still unclear, previous studies suggest that it may affect the localization of VAChT to synaptic vesicles.

Specificity

Other homologies: Human (95% sequence homology).
This antibody recognizes the C-terminus of Vesicular Acetylcholine Transporter (VAChT).

Immunogen

Epitope: C-terminus
KLH-conjugated linear peptide corresponding to the C-terminus of rat VAChT.

Application

Anti-Vesicular Acetylcholine Transporter (VAChT) Antibody detects level of Vesicular Acetylcholine Transporter (VAChT) & has been published & validated for use in IHC, ELISA.
Immunohistochemistry Analysis: A 1:1,000 dilution from a representative lot detected Vesicular Acetylcholine Transporter (VAChT) in rat midbrain tissue.
ELISA Analysis: A representative lot detected Vesicular Acetylcholine Transporter (VAChT) in a direct ELISA using a corresponding peptide.
Research Category
Neuroscience
Research Sub Category
Signaling Neuroscience

Quality

Evaluated by Immunohistochemistry Analysis.
Immunohistochemistry Analysis: A 1:1,000 dilution from a representative lot detected Vesicular Acetylcholine Transporter (VAChT) in mouse midbrain tissue.

Target description

57 kDa calculated

Physical form

Affinity purified
Purified goat polyclonal in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Analysis Note

Control
Vesicular Acetylcholine Transporter (VAChT) peptide

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

A genetically specified connectomics approach applied to long-range feeding regulatory circuits.
Atasoy, D; Betley, JN; Li, WP; Su, HH; Sertel, SM; Scheffer, LK; Simpson, JH; Fetter et al.
Nature Neuroscience null
Danyang Guo et al.
Metabolism: clinical and experimental, 107, 154232-154232 (2020-04-18)
Parasympathetic nerve (PN) signaling plays a crucial role in the maintenance of pancreatic β-cell volume density (Vβ). PN may be pathologically affected in diabetic polyneuropathy (DPN). However, the association between the reduction of PNs in islets and Vβ and the
Youssouf Cissé et al.
eNeuro, 5(4) (2018-09-19)
Acetylcholine (ACh) neurons in the pontomesencephalic tegmentum (PMT) are thought to play an important role in promoting cortical activation with waking (W) and paradoxical sleep [PS; or rapid eye movement (REM)], but have yet to be proven to do so
D B Jensen et al.
Scientific reports, 10(1), 893-893 (2020-01-23)
Intramuscular injections of botulinum toxin block pre-synaptic cholinergic release at neuromuscular junctions producing a temporary paralysis of affected motor units. There is increasing evidence, however, that the effects are not restricted to the periphery and can alter the central excitability
Marcin Bączyk et al.
The Journal of experimental medicine, 217(8) (2020-06-03)
Excessive excitation is hypothesized to cause motoneuron (MN) degeneration in amyotrophic lateral sclerosis (ALS), but actual proof of hyperexcitation in vivo is missing, and trials based on this concept have failed. We demonstrate, by in vivo single-MN electrophysiology, that, contrary

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service