Skip to Content
Merck

ABC97

Anti-Birc7/LIVIN Antibody

from rabbit, purified by affinity chromatography

Synonym(s):

Baculoviral IAP repeat-containing protein 7, Kidney inhibitor of apoptosis protein, KIAP, Livin, Melanoma inhibitor of apoptosis protein, ML-IAP, RING finger protein 50

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41
eCl@ss:
32160702
Conjugate:
unconjugated
Clone:
polyclonal
Application:
WB
Citations:
1
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


biological source

rabbit

Quality Segment

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

purified by

affinity chromatography

species reactivity

human

species reactivity (predicted by homology)

rhesus macaque (based on 100% sequence homology)

technique(s)

western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... BIRC7(79444)

General description

Livin, also known as “baculoviral IAP repeat-containing protein 7” (Birc7) and "KIAP," is a member of the IAP (inhibitors of apoptosis) family of proteins characterized by highly conserved baculoviral IAP repeats (BIR). In addition, Livin contains one RING-type zinc finger domain. Livin has been detected in the heart, placental, lung, lymph node, ovary, spleen, and in various types of solid tumors. It may suppress apoptosis induced by TNF, viral infection, drugs, and growth factor withdrawal, by inhibiting caspase activity (caspase 3, 7, and 9) and activating a MAPK/JUN pathway. Paradoxically, Livin may also function as a pro-apoptic factor in a truncated form (tLivin) that is cleaved by caspase 3 from the full-length protein. The pro-apoptotic effects of Livin may depend on its RING domain. Livin may play a role in non-Hodgkin lymphoma.
~39 kDa, ~32 kDa observed. Two isoforms at 32 kDa and 30 kDa are known to exist. This protein has been observed at ~39 kDa (alpha) and ~37 kDa (beta) due to post translational modification (Nachmias, B., et al. (2007). Apoptosis. 12(7):1129-1142.).

Immunogen

Epitope: BIR repeat region
KLH-conjugated linear peptide corresponding to the BIR repeat region of human Birc7/LIVIN.

Application

Anti-Birc7/LIVIN detects levels of Birc7/LIVIN proteins & has been published & validated for use in WB.
Research Category
Apoptosis & Cancer
Research Sub Category
Apoptosis - Additional

Biochem/physiol Actions

Other homologies: Rat, Mouse, and Bovine (93% sequence homology).
This antibody recognizes the BIR repeat region of Birc7/LIVIN.

Physical form

Affinity purified
Purified rabbit polyclonal serum in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Preparation Note

Stable for 1 year at 2-8°C from date of receipt.

Analysis Note

Control
Human spleen tissue lysate
Evaluated by Western Blot in human spleen tissue lysate.

Western Blot Analysis: 1 µg/mL of this antibody detected Birc7/LIVIN in 10 µg of human spleen tissue lysate.

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.


Still not finding the right product?

Try our Product Selector Tool to narrow your options


Storage Class

12 - Non Combustible Liquids

wgk

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library