Skip to Content
Merck
All Photos(1)

Key Documents

AB5986P

Sigma-Aldrich

Anti-Tyrosine Hydroxylase Antibody

Chemicon®, from rabbit

Synonym(s):

TH, Tyrosine Monooxygenase

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

rabbit

Quality Level

antibody form

affinity purified immunoglobulin

antibody product type

primary antibodies

clone

polyclonal

purified by

affinity chromatography

species reactivity

rat

manufacturer/tradename

Chemicon®

technique(s)

immunohistochemistry: suitable

UniProt accession no.

shipped in

dry ice

target post-translational modification

unmodified

Gene Information

human ... TH(7054)

Specificity

Tyrosine Hydroxylase. Immunohistochemical distribution throughout the brain shows that staining is restricted to those neurons known to contain the enzyme. The antibody has proven useful in staining catecholaminergic neurons. AB5986 stains these neurons intensely, including dendritic processes and fine nerve terminals.

Immunogen

Synthetic peptide corresponding to amino acids 32-47 of rat Tyrosine Hydroxylase.

Application

Detect Tyrosine Hydroxylase using this Anti-Tyrosine Hydroxylase Antibody validated for use in IH.
Immunohistochemistry: 0.5-1.0 μg/mL.

Optimal working dilutions must be determined by end user.
Research Category
Neuroscience
Research Sub Category
Neurotransmitters & Receptors

Neuronal & Glial Markers

Physical form

Affinity purified immunoglobulin.

Legal Information

CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Han-Ying Wang et al.
The Journal of physiology, 593(1), 161-180 (2015-01-06)
Noradrenaline (NA)-releasing neurons in the locus coeruleus (LC) provide NA to the forebrain and play important roles in regulating many brain functions. LC neurons are subject to tonic inhibition mediated by GABAB receptors (GABAB Rs) and that the extent of
Stephanie J Cragg et al.
The European journal of neuroscience, 20(7), 1788-1802 (2004-09-24)
The direct modulation of subthalamic nucleus (STN) neurons by dopamine (DA) neurons of the substantia nigra (SN) is controversial owing to the thick caliber and low density of DA axons in the STN. The abnormal activity of the STN in
Samantha K Smith et al.
ACS chemical neuroscience, 8(2), 272-280 (2016-12-17)
Cerebral blood flow ensures delivery of nutrients, such as glucose, to brain sites with increased metabolic demand. However, little is known about rapid glucose dynamics at discrete locations during neuronal activation in vivo. Acute exposure to many substances of abuse
Zhaoxia Luo et al.
Investigative ophthalmology & visual science, 61(11), 40-40 (2020-09-26)
The purpose of this study was to investigate the relationship between circadian rhythm and intraocular pressure (IOP), and to explore whether electrical stimulation of cervical sympathetic ganglia (SCG) can regulate IOP via neurotransmitter distribution around the Schlemm's canal (SC) in

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service